Chcete sa opäť prihlásiť?
Microplate centrifuge, PCR Plate Spinner
Dodávateľ: VWR Collection
The PCR Plate Spinner offers a personal sized centrifuge designed to spin down PCR plates.
  • Space saving design; quiet, simple operation
  • Easy plate loading system
  • Quickly spins down droplets and condensation
  • Use before and after thermal cycling to improve PCR yield
  • Cold room compatible
This unique centrifuge is equipped with a dual position vertical rotor that accepts a wide variety of PCR plates; 96-well, 384-well, skirted and non skirted plates are compatible with the Plate Spinner. The rotor features integral plate guides, ensuring that the plates stay vertical and do not tip inside the rotor.
Log in to see your contract pricing and availability.
  Zapamätať si ma na tomto zariadení
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
PCR plate spinner, EU-zástrčka 1 521-1648
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
ŠPECIFIKÁCIE
Model PCR Plate Spinner
Max. capacity 2 PCR platne
Max. rýchlosť (min-1) 2500
Max. RCF (×g) 500
Zdroj 230 V/50-60 Hz
ŠxH×V (mm) 190 × 2210 × 183
Hmotnosť (kg) 3
DOPLNKY
Rozšíriť všetko / Schovať všetko
Accessories
Opis Pk Číslo v katalógu VWR Množstvo
Adaptér pre stripy, skúmavky alebo poloplatne 2 521-1650
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo