Chcete sa opäť prihlásiť?
Aplikácie
Standards and Norms in water quality for drinking water, sewage waters, industrial waters and rivers.
Aplikácie
Water Quality
ISO 6059 Determination of Total Hardness ( calcium and magnesium) -- Titrimetric method with EDTA
ISO 6058 Determination of calcium -- Titrimetric method with EDTA
EN ISO 9963-1 Determination of Total Alcalinity & composite -- Part 1
EN ISO 9963-2 Determination of Alcalinity carbonate -- Part 2
ISO 9297 Determination of chlorides -- Titration with Silver nitrate and chromate as indicatorr (Mohr Méthod)
ISO 7393-1 Detrmination of free and total chlorine - Part 1: Titrimetric Mehod with N,N-diethylphenylene-1,4 diamine
ISO 7393-2 Detrmination of free and total chlorine -- Part 2: Colorimetric Method with N,N-diethylphenylene-1,4 diamine
ISO 15705 Determination of COD -- Micro method
ISO 6060 COD Determination
ISO/CD 25814 Dissolved oxygen determination -- electrochemical method with a D.O. probe
ISO/DIS 7887 Check and colour determination
ISO 7027 Turbidity determination
ISO 10359-1 Determination of fluorides -- Part 1: Electrochemical Method with ISE ( for drinking or low polluted waters )
ISO 5664 Determination of ammonium -- Method by distillation and titrimetry
ISO 10523 pH Determination
ISO 27888 Conductivity determination
ISO 11923 Determination of suspended materials by fibre glass filtration