Chcete sa opäť prihlásiť?
Celkový organický uhlík (TOC)
Celkový organický uhlík (TOC)
Total organic carbon (TOC) is the amount of carbon bound in an organic compound and is often used as a non-specific indicator of water quality.
  • Rapid digestion for C.O.D & T.O.C
  • Choice of photometers and spectrophotometers and ready to use T.O. C tubes and powder pillows.
  • 3 models:
    • TR 320: basic model for 12 cuvettes and 7 pre-stored temperature programmes
    • TR 420: accepts 24 cuvettes, in addition 8 programmes defined by the user
    • TR 620: accepts in addition 2 X 12 cuvettes with  2 temperature zones