Chcete sa opäť prihlásiť?
Integrator supplies and cables
Dodávateľ: Agilent Technologies
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Adapter, extends GPIB connector, 10834A 1 HEWL10834A-2310
Cable, 6890 to PC 9F/9F RS232 1 HEWLG1530-60600
Cable, 6890 to PC via RS 232-C, 9 female/25 male user card cable 1 HEWLG1530-60610
Cable, automatic liquid sampler, remote start/stop cable, 2 m, 9 male/9 female 1 HEWLG1530-60930
Cables, analog, to connect 3394/95A and 3396A/B integrator, general purpose 1 HEWL35900-60900
Cables, analog, to connect 3397A or 3396/95 series III integrator, for 5890 GC (non-INET) 1 HEWL35900-60610
Cables, analog, to connect 3397A or 3396/95 series III integrator, for 6890 series GC (non-INET) 1 HEWLG1530-60570
Cables, analog, to connect 3397A or 3396/95 series III integrator, for LC 1100/1200 series 1 HEWL35900-60750
Cables, analog, to connect 3397A or 3396/95 series III integrator, general purpose 1 HEWL35900-60630
Cables, remote, to connect 3394/95A and 3396A/B integrator, general purpose 1 HEWL35900-60920
Cables, remote, to connect 3397A or 3396/95 series III integrator, for LC 1100/1200 series/6890 series GC (non-INET) 1 HEWL03396-61010
Cables, remote, to connect 3397A or 3396/95 series III integrator, general purpose 1 HEWL03394-60540
Crossover ethertwist cable, 10 ft 1 HEWL5183-4649
External event control, cable, 8 pins/spade lugs 1 HEWLG1530-60590
General purpose cable GPIO, open end 1 HEWLG1103-61611
HPIB cable, 2 m, 10833B 1 HEWL10833B-2310
HPIB cable, 8 m, 10833G 1 HEWL10833G-2310
Ink cartridge, for agilent 3397A, 3396 series III, 3396A and B, 3395, 3394 1 HEWL5181-1220
PCI GPIB card for win 95/98/NT (82350A/B) 1 HEWLG1680-63715
Perforated paper, 8,5×11" rolls, inkjet, for agilent 3397A, 3396 series III, 3396A and B, 3395, 3394 4 Roll HEWL5183-2009
Perforated paper, 8,5×11" rolls, inkjet for agilent 3397A, 3396 series II, and 3396A/B/95/94 4 Roll HEWL5181-1219_P
Perforated paper, A4 size rolls, inkjet, for agilent 3397A, 3396 series III, 3396A and B, 3395, 3394 4 Roll HEWL5181-1255
Remote cable to 5890 1 HEWL35900-60700
Remote start cable for general use with lug 1 HEWL35900-60670
Remote Y cable, G1512/5890A to 3396C 1 HEWLG1512-60530
Replacement paper roll rod 1 HEWL03394-20500
Signal cable, general purpose analog output cable assembly, spade lugs/6 pins 1 HEWLG1530-60560
Thermal paper, blue print, for agilent 3390/92/93 4 Roll HEWL5080-8800
Z-fold paper, 8,5×11" rolls, for agilent 3397A, 3396 series II, and 3396A/B/95/94 5 HEWL5062-3561
Z-fold paper, black print, 8,5 wide, for agilent 338X/5830/3370 8 HEWL9270-0658
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo