Chcete sa opäť prihlásiť?
Stirrer blades
Dodávateľ: SciLabware
A range of PTFE coated stirrer heads designed to screw into the threads on SVL® glass stirrer shafts.
  • Head simply screws into the end of the shaft
  • Anchor and helix types have a PTFE coated metal core
Upozornení: Stirrers must be rotated in a clockwise direction to prevent unscrewing of the thread.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Typ Šírka Pk Číslo v katalógu VWR Množstvo
Collapsible stirrer blade, PTFE, for vessels with SVL 30 joint or 29/32 socket 70 mm 1 BIBB588-93
Paddle stirrer blades, PTFE, for SVL thermostatic reactors 70 mm 5 BIBB588-94
Anchor stirrer blade, PTFE, with stiffened metal core, for SVL reaction vessels 90 mm 1 BIBB588-95
Helix-6 blade stirrer, PTFE, with stiffened metal core, for SVL reaction vessels 80 mm 1 BIBB588-97
Anchor stirrer blades, PTFE, with stiffened metal core, for SVL reaction vessels 54 mm 5 BIBB588-98
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo