Chcete sa opäť prihlásiť?
Micro-V vials, CrossLab
Dodávateľ: Agilent Technologies
CrossLab
These vials are made from first hydrolytic glass type 1 and are available with screw, crimp or snap tops.
  • Wide opening with low residual volume
  • Compatible with 11 mm crimp closures
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Typ Kapacita Ø×H Pk Číslo v katalógu VWR Množstvo
Amber glass, screw top 1,50 ml 12×32 mm 100 HEWL8010-0019
Clear glass, crimp top 1,50 ml 12×32 mm 100 HEWL8010-0004
Amber glass, crimp top 1,50 ml 12×32 mm 100 HEWL8010-0005
Clear glass, snap top 1,50 ml 12×32 mm 100 HEWL8010-0027
Amber glass, snap top 1,50 ml 12×32 mm 100 HEWL8010-0028
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo