Chcete sa opäť prihlásiť?
HPLC columns, NUCLEODUR® C8 gravity
Dodávateľ: MACHEREY-NAGEL
NUCLEOSIL® NUCLEODUR®
These columns are particularly suitable for the separation of basic and other ionisable analytes. Even strongly basic pharmaceuticals like amitriptyline are eluted without tailing under isocratic conditions.
  • Superior base deactivation
  • Suitable for LC/MS and HPLC at pH extremes (pH 1 - 11)
  • Overall sophisticated analytical separations that is ideal for method development
Recommended application in pharmaceuticals in areas such as analgesics, anti-inflammatory drugs, antidepressants; herbicides; phytopharmaceuticals; immunosuppressants.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
VP guard columns are used for preparative VarioPrep columns with the following guard column holders: 8,0×10 mm (Int.Ø×L) guard column for 8,0 and 10,0 mm (Int.Ø) preparative column with holder MANA718251. 16,0×10 mm (Int.Ø×L) guard column for 16,0 and 21,0 mm (Int.Ø) preparative column with holder MANA718256. 32,0×15 mm (Int.Ø×L) guard column for 32,0 and 40,0 mm (Int.Ø) preparative column with holder MANA718253. 50,0×15 mm (Int.Ø×L) guard column for ≥50,0 mm (Int.Ø) preparative column with holder MANA718255.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C8 Gravity 110 Å 1,8 µm 2,0×30 mm 1 MANA760756.20
Analytical C8 Gravity 110 Å 1,8 µm 2,0×50 mm 1 MANA760755.20
Analytical C8 Gravity 110 Å 3 µm 2,0×50 mm 1 MANA760653.20
Analytical C8 Gravity 110 Å 5 µm 2,0×50 mm 1 MANA760750.20
Analytical C8 Gravity 110 Å 1,8 µm 2,0×75 mm 1 MANA760760.20
Analytical C8 Gravity 110 Å 3 µm 2,0×75 mm 1 MANA760655.20
Analytical C8 Gravity 110 Å 1,8 µm 2,0×100 mm 1 MANA760757.20
Analytical C8 Gravity 110 Å 3 µm 2,0×100 mm 1 MANA760656.20
Analytical C8 Gravity 110 Å 5 µm 2,0×100 mm 1 MANA760754.20
Analytical C8 Gravity 110 Å 3 µm 2,0×125 mm 1 MANA760657.20
Analytical C8 Gravity 110 Å 5 µm 2,0×125 mm 1 MANA760751.20
Analytical C8 Gravity 110 Å 1,8 µm 2,0×150 mm 1 MANA760759.20
Analytical C8 Gravity 110 Å 3 µm 2,0×150 mm 1 MANA760658.20
Analytical C8 Gravity 110 Å 5 µm 2,0×150 mm 1 MANA760752.20
Analytical C8 Gravity 110 Å 3 µm 2,0×250 mm 1 MANA760659.20
Analytical C8 Gravity 110 Å 5 µm 2,0×250 mm 1 MANA760753.20
Analytical C8 Gravity 110 Å 1,8 µm 3,0×30 mm 1 MANA760756.30
Analytical C8 Gravity 110 Å 1,8 µm 3,0×50 mm 1 MANA760755.30
Analytical C8 Gravity 110 Å 3 µm 3,0×50 mm 1 MANA760653.30
Analytical C8 Gravity 110 Å 5 µm 3,0×50 mm 1 MANA760750.30
Analytical C8 Gravity 110 Å 3 µm 3,0×75 mm 1 MANA760655.30
Analytical C8 Gravity 110 Å 1,8 µm 3,0×100 mm 1 MANA760757.30
Analytical C8 Gravity 110 Å 3 µm 3,0×100 mm 1 MANA760656.30
Analytical C8 Gravity 110 Å 5 µm 3,0×100 mm 1 MANA760754.30
Analytical C8 Gravity 110 Å 3 µm 3,0×125 mm 1 MANA760657.30
Analytical C8 Gravity 110 Å 5 µm 3,0×125 mm 1 MANA760751.30
Analytical C8 Gravity 110 Å 3 µm 3,0×150 mm 1 MANA760658.30
Analytical C8 Gravity 110 Å 5 µm 3,0×150 mm 1 MANA760752.30
Analytical C8 Gravity 110 Å 3 µm 3,0×250 mm 1 MANA760659.30
Analytical C8 Gravity 110 Å 5 µm 3,0×250 mm 1 MANA760753.30
Analytical C8 Gravity 110 Å 1,8 µm 4,0×30 mm 1 MANA760756.40
Analytical C8 Gravity 110 Å 1,8 µm 4,0×50 mm 1 MANA760755.40
Analytical C8 Gravity 110 Å 3 µm 4,0×50 mm 1 MANA760653.40
Analytical C8 Gravity 110 Å 5 µm 4,0×50 mm 1 MANA760750.40
Analytical C8 Gravity 110 Å 3 µm 4,0×75 mm 1 MANA760655.40
Analytical C8 Gravity 110 Å 1,8 µm 4,0×100 mm 1 MANA760757.40
Analytical C8 Gravity 110 Å 3 µm 4,0×100 mm 1 MANA760656.40
Analytical C8 Gravity 110 Å 5 µm 4,0×100 mm 1 MANA760754.40
Analytical C8 Gravity 110 Å 3 µm 4,0×125 mm 1 MANA760657.40
Analytical C8 Gravity 110 Å 5 µm 4,0×125 mm 1 MANA760751.40
Analytical C8 Gravity 110 Å 3 µm 4,0×150 mm 1 MANA760658.40
Analytical C8 Gravity 110 Å 5 µm 4,0×150 mm 1 MANA760752.40
Analytical C8 Gravity 110 Å 3 µm 4,0×250 mm 1 MANA760659.40
Analytical C8 Gravity 110 Å 5 µm 4,0×250 mm 1 MANA760753.40
Analytical C8 Gravity 110 Å 1,8 µm 4,6×30 mm 1 MANA760756.46
Analytical C8 Gravity 110 Å 1,8 µm 4,6×50 mm 1 MANA760755.46
Analytical C8 Gravity 110 Å 3 µm 4,6×50 mm 1 MANA760653.46
Analytical C8 Gravity 110 Å 5 µm 4,6×50 mm 1 MANA760750.46
Analytical C8 Gravity 110 Å 3 µm 4,6×75 mm 1 MANA760655.46
Analytical C8 Gravity 110 Å 5 µm 4,6×75 mm 1 MANA760749.46
Analytical C8 Gravity 110 Å 1,8 µm 4,6×100 mm 1 MANA760757.46
Analytical C8 Gravity 110 Å 3 µm 4,6×100 mm 1 MANA760656.46
Analytical C8 Gravity 110 Å 5 µm 4,6×100 mm 1 MANA760754.46
Analytical C8 Gravity 110 Å 3 µm 4,6×125 mm 1 MANA760657.46
Analytical C8 Gravity 110 Å 5 µm 4,6×125 mm 1 MANA760751.46
Analytical C8 Gravity 110 Å 3 µm 4,6×150 mm 1 MANA760658.46
Analytical C8 Gravity 110 Å 5 µm 4,6×150 mm 1 MANA760752.46
Analytical C8 Gravity 110 Å 3 µm 4,6×250 mm 1 MANA760659.46
Analytical C8 Gravity 110 Å 5 µm 4,6×250 mm 1 MANA760753.46
Preparative columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Preparative C8 Gravity 110 Å 5 µm 10,0×50 mm 1 MANA762081.100
Preparative C8 Gravity 110 Å 5 µm 10,0×125 mm 1 MANA762071.100
Preparative C8 Gravity 110 Å 5 µm 10,0×250 mm 1 MANA762070.100
Preparative C8 Gravity 110 Å 5 µm 21,0×150 mm 1 MANA762082.210
Preparative C8 Gravity 110 Å 5 µm 21,0×250 mm 1 MANA762070.210
Guard columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column C8 Gravity 110 Å 1,8 µm 2,0×4 mm 3 MANA761905.20
Guard column C8 Gravity 110 Å 3 µm 2,0×4 mm 3 MANA761906.20
Guard column C8 Gravity 110 Å 5 µm 2,0×4 mm 3 MANA761907.20
Guard column C8 Gravity 110 Å 5 µm 3,0×4 mm 3 MANA761907.30
Guard column C8 Gravity 110 Å 1,8 µm 3,0×4 mm 3 MANA761905.30
Guard column C8 Gravity 110 Å 3 µm 3,0×4 mm 3 MANA761906.30
VP guard columns
Column format Phase Veľkosť pórov Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column C8 Gravity 110 Å 8,0×10 mm 1 MANA762097.80
Guard column C8 Gravity 110 Å 16,0×10 mm 2 MANA762097.160
Guard column holders for VP columns
Opis Pk Číslo v katalógu VWR Množstvo
VarioPrep guard column holder, for VarioPrep columns 8 - 10 mm I-Ø 1 MANA718251
VP guard column holder 16 mm for VarioPrep columns with 16 and 21 mm I-Ø 1 MANA718253
VP guard column holder 32 mm for VarioPrep columns with 32 and 40 mm I-Ø 1 MANA718255
VP guard column holder 50 mm for VarioPrep columns with ≥50 mm I-Ø 1 MANA718256
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Carbon load 11%
Endcapped Yes
pH range 1 - 11
USP code L7
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo