Tlačiť
Pipette tips, racked, for Pos-D™ pipettes
Dodávateľ: Mettler-Toledo
RAINC-25 RAINC-250 RAIN17008607 RAIN17008606 RAINC-1000 RAIN17008605 RAIN17008604 RAINC-50 RAIN17008609 RAIN17008608 RAINC-10
Pipette tips, racked, for Pos-D™ pipettes
Špičky pipetovací
These tips are designed for Pos-D™ positive displacement pipettes. A disposable piston, which moves within a plastic capillary, makes direct contact with the liquid. The positive wiping action of the piston against the capillary wall assures complete dispensing without droplets. These tips eliminate cross-contamination from aerosols, samples or pipette.