Tlačiť
Sample bottles, borosilicate glass, BHROTEST®
Dodávateľ: Behr
behrotest®
719-1303 719-1302 719-1301
Sample bottles, borosilicate glass, BHROTEST®
Láhve Láhve skleněné
Borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Glass sampling bottle with conical ground glass stopper.
Certifikácie: Conforming to DIN 12036.