Chcete sa opäť prihlásiť?
Scalpel handle with blades
Dodávateľ: MURRAY, S & CO LTD
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Handles
Typ Pre Opis Pk Číslo v katalógu VWR Množstvo
No. 3 Blades (No. 10/10a/11/12/15) Scalpel handle 1 MURR1802/05
No. 4 Blades (No. 20/21/22/22a/23/24/25/26/27) Scalpel handle 1 MURR1802/15
No. 7 Blades (No. 10/10a/11/12/15) Scalpel handle 1 MURR1802/25
Blades
Typ Pre Opis Pk Číslo v katalógu VWR Množstvo
No. 10 Handles No. 3/3L/5 Scalpel blade, carbon, for general surgery 1 MURR1803/05
No. 10A Handles No. 3/3L/5 Scalpel blade, carbon, for general surgery 1 MURR1803/10
No. 11 Handles No. 3/3L/5 Scalpel blade, carbon, for general surgery 1 MURR1803/15
No. 12 Handles No. 3/3L/5 Scalpel blade, carbon, for general surgery 1 MURR1803/20
No. 15 Handles No. 3/3L/5 Scalpel blade, carbon, for general surgery 1 MURR1803/25
No. 20 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/30
No. 21 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/35
No. 22 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/40
No. 22A Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/45
No. 23 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/50
No. 24 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/55
No. 25 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/60
No. 26 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/65
No. 27 Handles No. 4/4L Scalpel blade, carbon, for general surgery 1 MURR1803/70
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo