Tlačiť
Aluminium blocks, Multi-Blok®
Dodávateľ: Thermo Scientific
BARN2066QEA 0 EUR
BARN2066Q BARN2072Q BARN2083Q BARN2073Q BARN2070Q
Aluminium blocks, Multi-Blok®
Made from anodised aluminium, close contact of tubes to block walls provides excellent heat transfer.
  • Blocks have a separate hole to accommodate a thermometer if required
  • Block puller included with all models