Chcete sa opäť prihlásiť?
Gel permeation chromatography columns, NUCLEOGEL® GPC
Dodávateľ: MACHEREY-NAGEL
Polymer based phases for size exclusion chromatography (SEC) or gel permeation chromatography (GPC) of water insoluble substances.
  • Highly crosslinked macroporous, spherical polystyrene divinylbenzene
  • Good mechanical stability
  • Eluent in column toluene
The Valco type guard column will be connected with Valco type analytical column by a short capillary.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Opis Column format Particle size Int.Ø×L Exclusion limit Pk Číslo v katalógu VWR Množstvo
NUCLEOGEL® GPC 50-5 Analytical 5 µm 7,7×300 mm 2 kD 1 MANA719402
NUCLEOGEL® GPC 100-5 Analytical 5 µm 7,7×300 mm 4 kD 1 MANA719403
NUCLEOGEL® GPC 500-5 Analytical 5 µm 7,7×300 mm 25 kD 1 MANA719404
NUCLEOGEL® GPC 103-5 Analytical 5 µm 7,7×300 mm 60 kD 1 MANA719405
NUCLEOGEL® GPC 104-5 Analytical 5 µm 7,7×300 mm 500 kD 1 MANA719406
NUCLEOGEL® GPC 50-10 Analytical 10 µm 7,7×300 mm 2 kD 1 MANA719410
NUCLEOGEL® GPC 100-10 Analytical 10 µm 7,7×300 mm 4 kD 1 MANA719411
NUCLEOGEL® GPC 500-10 Analytical 10 µm 7,7×300 mm 25 kD 1 MANA719412
NUCLEOGEL® GPC 103-10 Analytical 10 µm 7,7×300 mm 60 kD 1 MANA719413
NUCLEOGEL® GPC 104-10 Analytical 10 µm 7,7×300 mm 500 kD 1 MANA719414
Guard columns
Opis Column format Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
NUCLEOGEL® GPC 5P Guard column 5 µm 7,7×50 mm 1 MANA719409
NUCLEOGEL® GPC 10P Guard column 10 µm 7,7×50 mm 1 MANA719418
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo