Chcete sa opäť prihlásiť?
Preparative HPLC columns, PLRP-S
Dodávateľ: Agilent Technologies
The PLRP-S media, rigid poly(styrene/divinylbenzene) particles, are available in a range of pore sizes for small molecule, synthetic biomolecule and macromolecule purification. Their thermal and chemical stability makes them ideal for purifications that require extreme conditions for sample preparation, compound elution and column regeneration
  • Discovery stage to multi-kg cGMP production reduces method development time
  • Excellent chemical stability for separations, sanitation and regeneration increases selectivity and column lifetime
  • Single batch packing of multiple columns reduces system down time and validation costs
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Prep to process
Column format Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Preparative 100 Å 8 µm 25×150 mm 1 VARIPL1212-3800
Preparative 100 Å 8 µm 25×300 mm 1 VARIPL1212-6800
Preparative 100 Å 8 µm 50×150 mm 1 VARIPL1712-3800
Preparative 100 Å 8 µm 50×300 mm 1 VARIPL1712-6800
Preparative 100 Å 10 µm 25×150 mm 1 VARIPL1212-3100
Preparative 100 Å 10 µm 25×300 mm 1 VARIPL1212-6100
Preparative 100 Å 10 µm 50×150 mm 1 VARIPL1712-3100
Preparative 100 Å 10 µm 100×300 mm 1 VARIPL1812-6100
Preparative 100 Å 10 - 15 µm 25×300 mm 1 VARIPL1212-6400
Preparative 100 Å 10 - 15 µm 50×150 mm 1 VARIPL1712-3400
Preparative 100 Å 10 - 15 µm 100×300 mm 1 VARIPL1812-6400
Preparative 100 Å 15 - 20 µm 25×300 mm 1 VARIPL1212-6200
Preparative 100 Å 15 - 20 µm 50×150 mm 1 VARIPL1712-3200
Preparative 100 Å 15 - 20 µm 100×300 mm 1 VARIPL1812-6200
Preparative 300 Å 8 µm 25×150 mm 1 VARIPL1212-3801
Preparative 300 Å 8 µm 25×300 mm 1 VARIPL1212-6801
Preparative 300 Å 8 µm 50×150 mm 1 VARIPL1712-3801
Preparative 300 Å 8 µm 50×300 mm 1 VARIPL1712-6801
Preparative 300 Å 10 µm 25×150 mm 1 VARIPL1212-3101
Preparative 300 Å 10 µm 25×300 mm 1 VARIPL1212-6101
Preparative 300 Å 10 µm 50×150 mm 1 VARIPL1712-3101
Preparative 300 Å 10 µm 100×300 mm 1 VARIPL1812-6101
Preparative 300 Å 10 - 15 µm 25×300 mm 1 VARIPL1212-6401
Preparative 300 Å 10 - 15 µm 50×150 mm 1 VARIPL1712-3401
Preparative 300 Å 10 - 15 µm 100×300 mm 1 VARIPL1812-6401
Preparative 300 Å 15 - 20 µm 25×300 mm 1 VARIPL1212-6201
Preparative 300 Å 15 - 20 µm 50×150 mm 1 VARIPL1712-3201
Preparative 300 Å 15 - 20 µm 100×300 mm 1 VARIPL1812-6201
Preparative 1000 Å 10 µm 25×50 mm 1 VARIPL1212-1102
Preparative 1000 Å 10 µm 25×150 mm 1 VARIPL1212-3102
Preparative 1000 Å 10 µm 50×150 mm 1 VARIPL1712-3102
Preparative 1000 Å 10 µm 100×300 mm 1 VARIPL1812-2102
Preparative 1000 Å 30 µm 25×150 mm 1 VARIPL1212-3702
Preparative 1000 Å 30 µm 50×150 mm 1 VARIPL1712-3702
Preparative 1000 Å 30 µm 100×300 mm 1 VARIPL1812-3102
Preparative 4000 Å 10 µm 25×50 mm 1 VARIPL1212-1103
Preparative 4000 Å 10 µm 25×150 mm 1 VARIPL1212-3103
Preparative 4000 Å 10 µm 50×150 mm 1 VARIPL1712-3103
Preparative 4000 Å 10 µm 100×300 mm 1 VARIPL1812-2103
Preparative 4000 Å 30 µm 25×150 mm 1 VARIPL1212-3703
Preparative 4000 Å 30 µm 50×150 mm 1 VARIPL1712-3703
Preparative 4000 Å 30 µm 100×300 mm 1 VARIPL1812-3103
Method development
Column format Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical 100 Å 10 µm 4,6×150 mm 1 VARIPL1512-3100
Analytical 100 Å 10 µm 4,6×250 mm 1 VARIPL1512-5100
Preparative 100 Å 10 µm 50×300 mm 1 VARIPL1712-6100
Analytical 100 Å 10 - 15 µm 4,6×150 mm 1 VARIPL1512-3400
Analytical 100 Å 10 - 15 µm 4,6×250 mm 1 VARIPL1512-5400
Preparative 100 Å 10 - 15 µm 50×300 mm 1 VARIPL1712-6400
Analytical 100 Å 15 - 20 µm 4,6×150 mm 1 VARIPL1512-3200
Analytical 100 Å 15 - 20 µm 4,6×250 mm 1 VARIPL1512-5200
Preparative 100 Å 15 - 20 µm 50×300 mm 1 VARIPL1712-6200
Analytical 300 Å 10 µm 4,6×150 mm 1 VARIPL1512-3101
Analytical 300 Å 10 µm 4,6×250 mm 1 VARIPL1512-5101
Preparative 300 Å 10 µm 50×300 mm 1 VARIPL1712-6101
Analytical 300 Å 10 - 15 µm 4,6×150 mm 1 VARIPL1512-3401
Analytical 300 Å 10 - 15 µm 4,6×250 mm 1 VARIPL1512-5401
Preparative 300 Å 10 - 15 µm 50×300 mm 1 VARIPL1712-6401
Analytical 300 Å 15 - 20 µm 4,6×150 mm 1 VARIPL1512-3201
Analytical 300 Å 15 - 20 µm 4,6×250 mm 1 VARIPL1512-5201
Preparative 300 Å 15 - 20 µm 50×300 mm 1 VARIPL1712-6201
Analytical 1000 Å 10 µm 4,6×150 mm 1 VARIPL1512-3102
Analytical 1000 Å 10 µm 4,6×250 mm 1 VARIPL1512-5102
Analytical 1000 Å 30 µm 4,6×150 mm 1 VARIPL1512-3702
Analytical 1000 Å 30 µm 4,6×250 mm 1 VARIPL1512-5702
Analytical 4000 Å 10 µm 4,6×150 mm 1 VARIPL1512-3103
Analytical 4000 Å 10 µm 4,6×250 mm 1 VARIPL1512-5103
Analytical 4000 Å 30 µm 4,6×150 mm 1 VARIPL1512-3703
Analytical 4000 Å 30 µm 4,6×250 mm 1 VARIPL1512-5703
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
pH range 1-14
Temperature limit 200 ºC
USP code L21
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo