Chcete sa opäť prihlásiť?
Dressing jars
Dodávateľ: S Murray & Co
Dressing jars, made of moulded glass, round.
  • Good chemical resistance
  • Lids must be ordered seperately
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Výška Vonkajší Ø. Pk Číslo v katalógu VWR Množstvo
100 mm 100 mm 1 MURRG500/01
125 mm 125 mm 1 MURRG500/04
150 mm 100 mm 1 MURRG500/02
150 mm 150 mm 1 MURRG500/05
200 mm 100 mm 1 MURRG500/03
200 mm 150 mm 1 MURRG500/06
200 mm 200 mm 1 MURRG500/08
250 mm 150 mm 1 MURRG500/07
250 mm 200 mm 1 MURRG500/09
300 mm 200 mm 1 MURRG500/10
Lids
Opis Pk Číslo v katalógu VWR Množstvo
Lids, moulded glass, circular, for jars of O.Ø 100 mm 1 MURRG501/01
Lids, moulded glass, circular, for jars of O.Ø 125 mm 1 MURRG501/02
Lids, moulded glass, circular, for jars of O.Ø 150 mm 1 MURRG501/03
Lids, stainless steel, circular with plain rim, for jars of O.Ø 100 mm 1 MURRG502/01
Lids, stainless steel, circular with plain rim, for jars of O.Ø 125 mm 1 MURRG502/02
Lids, stainless steel, circular with plain rim, for jars of O.Ø 150 mm 1 MURRG502/03
Lids, stainless steel, circular with plain rim, for jars of O.Ø 200 mm 1 MURRG502/04
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo