Chcete sa opäť prihlásiť?
Preparative columns, PLgel™
Dodávateľ: Agilent Technologies
These preparative columns is ideal to fractionate polymers, isolate components in a polymer formulation or simplify mixtures of relatively small molecules in complex matrices.
  • Excellent column efficiency provides optimum resolution
  • High loading can isolate milli grams amounts for further study
  • Over 10 times scale up permits efficient quantification
It is packed with the same rigid, high performance media as the analytical columns so that mixtures of materials are easily separated on the basis of size, preferably in a low boiling organic solvent. They are then collected as a series of discrete fractions and isolated by simple evaporation of the solvent.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Column format Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
PLgel™, MW ranges upto 2000 Preparative 50 Å 10 µm 25×300 mm 1 VARIPL1210-6115
PLgel™, MW ranges upto 4000 Preparative 100 Å 10 µm 25×300 mm 1 VARIPL1210-6120
PLgel™, MW ranges 500 - 30000 Preparative 500 Å 10 µm 25×300 mm 1 VARIPL1210-6125
PLgel™, MW ranges 500 - 60000 Preparative 1000 Å 10 µm 25×300 mm 1 VARIPL1210-6130
PLgel™, MW ranges 10000 - 600000 Preparative 10000 Å 10 µm 25×300 mm 1 VARIPL1210-6140
PLgel™, MW ranges 60000 - 2000000 Preparative 100000 Å 10 µm 25×300 mm 1 VARIPL1210-6150
PLgel™, MW ranges 600000 - 10000000 Preparative 1000000 Å 10 µm 25×300 mm 1 VARIPL1210-6160
PLgel™, Mixed-D, MW ranges 200 - 400000 Preparative - 10 µm 25×300 mm 1 VARIPL1210-6104
PLgel™, Mixed-B, MW ranges 500 - 10000000 Preparative - 10 µm 25×300 mm 1 VARIPL1210-6100
PLgel™, Prep guard Guard column - - µm 25×25 mm 1 VARIPL1210-1120
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo