Chcete sa opäť prihlásiť?
Accessories for BOD measurement, Behrotest®
Dodávateľ: Behr
behrotest®
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Dilution units
Opis Pk Číslo v katalógu VWR Množstvo
BSB 4A dilution unit is semi-automatic with rinse mechanism 1 715-5162
BSB 2 diltution unit, used for bubble-free supply of dilution water 1 715-5161
BOD bottles
Opis Pk Číslo v katalógu VWR Množstvo
Bottles, Karlsruhe type with 250 ml capacity and 30 mm finger grip length 1 BEHRB00217779
Bottles, Karlsruhe type with 250 ml capacity and 60 mm finger grip length 1 715-3250
Bottles, Karlsruhe type with 100 ml capacity and 30 mm finger grip length 1 715-5115
Bottles, Karlsruhe type with 100 ml capacity and 60 mm finger grip length 1 BEHRB00217784
Dilution containers, without coil
Opis Pk Číslo v katalógu VWR Množstvo
Containers VD 30, blue, with aeration heads and air pressure connection, volume 30 l 1 715-5111
Containers VD 60, blue, with aeration heads and air pressure connection, volume 60 l 1 715-5112
Containers VD 120, blue, with aeration heads and air pressure connection, volume 120 l 1 715-5113
Dilution containers, with coil
Opis Pk Číslo v katalógu VWR Množstvo
Containers VDT 30, as VD 30 but with internal cooling/warming coil and connection for thermostat-circulation pump, volume 30 l 1 715-5121
Containers VDT 60, as VD 60 but with internal cooling/warming coil and connection for thermostat-circulation pump, volume 60 l 1 715-5122
Containers VDT 120, as VD 120, but with internal cooling/warming coil and connection for thermostat-circulation pump, volume 120 l 1 715-5123
Diaphragm pumps
Opis Pk Číslo v katalógu VWR Množstvo
Diaphragm pump MP 300, with maximum flow rate (300 l/hr), 4 W power consumption 1 715-5141
Diaphragm pump MP 500, with maximum flow rate (500 l/hr), 8 W power consumption 1 715-5142
Thermostatic circulator
Opis Pk Číslo v katalógu VWR Množstvo
Thermostatic circulation pumps maintains the BOD dilution water 20 °C by active cooling or heating, EU-plug 1 715-5131
Magnetic stirrer
Opis Pk Číslo v katalógu VWR Množstvo
Magnetic stirrer, for 250 ml bottles with external dimension 80×100×150 mm 1 715-5151
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo