Chcete sa opäť prihlásiť?
Flash chromatography glass columns and kits
Dodávateľ: MACHEREY-NAGEL
These low pressure flash chromatography columns and kits are suitable for the separation of compounds up to gram levels.
  • Economic low-tech method for the synthesis laboratory
Informace o dodávce: Flash chromatography columns are delivered complete with adapter and PTFE tap, fitted with a polyethylene net to protect against bursting.
Flash chromatography kit I, consists of 1 glass column 20×400 mm, one 1 litre eluent reservoir, 100 g silica 60 (40 - 63 µm), sea sand, silanised glass fiber wadding, 1 m PTFE tubing.
Flash chromatography kit II, consists of 1 glass column 40×450 mm, one 2 litre eluent reservoir, 100 g silica 60 (40 - 63 µm), sea sand, silanised glass fiber wadding, 1 m PTFE tubing.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Columns
Column hardware Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Glass 40 - 63 µm 20×200 mm 1 KIT MANA727400
Glass 40 - 63 µm 20×400 mm 1 KIT MANA727401
Glass 40 - 63 µm 25×200 mm 1 KIT MANA727402
Glass 40 - 63 µm 25×400 mm 1 KIT MANA727403
Glass 40 - 63 µm 30×300 mm 1 KIT MANA727404
Glass 40 - 63 µm 30×400 mm 1 KIT MANA727405
Glass 40 - 63 µm 40×300 mm 1 KIT MANA727406
Glass 40 - 63 µm 40×450 mm 1 KIT MANA727407
Kits
Column hardware Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Glass 40 - 63 µm 20×400 mm 1 KIT MANA727450
Glass 40 - 63 µm 40×450 mm 1 KIT MANA727451
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
DOPLNKY
Rozšíriť všetko / Schovať všetko
Accessories
Opis Pk Číslo v katalógu VWR Množstvo
Eluent reservoir 1 l with adapter, covered with a protective plastic sleeve for burst protection 1 KIT MANA727420
Eluent reservoir 2 l with adapter, covered with a protective plastic sleeve for burst protection 1 KIT MANA727421
Pressure gauge for controlling flow rates 1 MANA727422
Sea sand, acid washed and calcined 1.000 g MANA727423
Glass fiber wadding, silanized, very fine fibers 25 g MANA718002
PTFE tubing, 3 mm OD, 2 mm ID, 1 m length 1 m MANA727424
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo