Chcete sa opäť prihlásiť?
Rockers, Vari-Mix™ and Speci-Mix™
Dodávateľ: Thermo Scientific
Vari-Mix™
Vari-Mix™ and Speci-Mix™ Test Tube Rockers have smooth rocking action helps to obtain the uniform specimen suspension. These compact and precision-controlled platform mixers can hold tubes of Ø 10 to 30 mm and are ideal for blood collection tubes.
  • Choose from fixed or adjustable speeds
  • Autoclavable white silicone rubber pad enhances viewing
Reversible pad provides one side for 110 mm long capped tubes and other side for longer tubes. These rockers can be used in a noncondensing incubator or cold room with ambient operating conditions of 17 ° to 27 °C and 20 to 80 % RH noncondensing.
Informace o dodávce: Includes white silicon rubber friction grip pad for holding tubes in place.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Vari-Mix™ Test tube rocker 1 BARNM48720-33Q
Speci-Mix™ Test tube rocker 1 BARNM71010-33Q
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Model M48720-33 M71010-33
Kapacita 10 - 20 mm Ø tubes:16
22 - 30 mm Ø tubes: 7
10 - 20 mm Ø tubes: 8
22 - 30 mm Ø tubes: 3
Pohyb miešania Rocking
Rozsah otáčok (min-1) 2 − 20 18
Angle (°) 1 − 48, adjustable 48, fixed
Max. zaťaženie (kg) 0,56
ŠxH×V (mm) 381×139×107 191×139×107
Hmotnosť (kg) 2,6 1,4
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo