Chcete sa opäť prihlásiť?
Cell culture flasks, treated, Nunc™
Dodávateľ: Thermo Scientific
Nunc®
PS, sterile
  • Quick volume estimation and sample identification; includes bold printed graduationsfrom 100 to 500 mL, in 50 mL graduations and large writing patch
  • Maintains optimal sterility during growth and passage; filter cap incorporates a 0.2 µm pore size hydrophobic membrane that allows gas exchange while maintaining sterility
  • Easy to stack; prominent stacking feet on upper surface facilitate reliable stacking in incubators and under your culture hood
This total volume flasks provides a large culture area of single-layer flasks and is cell culture treated to promote cell attachment and expanded growth. Durable construction, friendly design; blow-molded, one-piece design with straight neck and molded grip notches that make this 1,900mL flask easy to hold. Non-pyrogenic.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Uzáver Rastová plocha Hrdlo Pk Číslo v katalógu VWR Množstvo
1900 ml - 300 cm² straight 12 734-2609
1900 ml Vent/close 300 cm² straight 12 734-2608
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo