Chcete sa opäť prihlásiť?
Ion exchange chromatography columns, PL-SAX and PL-SCX
Dodávateľ: Agilent Technologies
These rigid, strong ion exchange materials are extremely hydrophilic that is ideal for purification of biomolecules. The PL-SAX and PL-SCX materials are totally polymeric and are chemically and thermally stable over a full range of HPLC conditions.
  • Ion exchange purifications over a wider pH range extend applications
  • HPLC flow rates and rapid equilibration reduce purification cycle times
  • Large pore size for improved mass transfer delivers high speed, high resolution purifications
  • The 1000 Å pore size is for high capacity purifications
  • The 4000 Å with improved mass transfer are intended for large biomolecules and high-speed, high-resolution purifications
Thermal stability enables the use of denaturing conditions and stabilizing/solubilizing agents for the purification of target compounds that may associate or degrade under the purification conditions, such as the purification of synthetic oligonucleotides
with self-complementary sequences.
Both the 1000 Å and 4000 Å wide-pore materials are mechanically stable and robust and can be operated over a wide range of linear velocities with fast loading of dilute solutions and wash cycles. HPLC flow rates and rapid equilibration reduces purification cycle times.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
PL-SAX
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical PL-SAX 1000 Å 8 µm 2,1×150 mm 1 VARIPL1951-3802
Analytical PL-SAX 1000 Å 8 µm 7,5×50 mm 1 VARIPL1151-1802
Analytical PL-SAX 1000 Å 10 µm 4,6×150 mm 1 VARIPL1551-3102
Analytical PL-SAX 1000 Å 10 µm 4,6×250 mm 1 VARIPL1551-5102
Analytical PL-SAX 1000 Å 30 µm 4,6×150 mm 1 VARIPL1551-3702
Analytical PL-SAX 1000 Å 30 µm 4,6×250 mm 1 VARIPL1551-5702
Analytical PL-SAX 4000 Å 8 µm 4,6×150 mm 1 VARIPL1551-3803
Analytical PL-SAX 4000 Å 8 µm 7,5×50 mm 1 VARIPL1151-1803
Analytical PL-SAX 4000 Å 8 µm 7,5×150 mm 1 VARIPL1151-3803
Analytical PL-SAX 4000 Å 10 µm 4,6×150 mm 1 VARIPL1551-3103
Analytical PL-SAX 4000 Å 10 µm 4,6×250 mm 1 VARIPL1551-5103
Analytical PL-SAX 4000 Å 30 µm 4,6×250 mm 1 VARIPL1551-5703
Preparative PL-SAX 1000 Å 10 µm 25×50 mm 1 VARIPL1251-1102
Preparative PL-SAX 1000 Å 30 µm 50×150 mm 1 VARIPL1751-3702
Preparative PL-SAX 4000 Å 30 µm 25×150 mm 1 VARIPL1251-3703
Preparative PL-SAX 4000 Å 10 µm 25×150 mm 1 VARIPL1251-3103
Preparative PL-SAX 4000 Å 10 µm 25×50 mm 1 VARIPL1251-1103
Preparative PL-SAX 4000 Å 10 µm 50×150 mm 1 VARIPL1751-3103
Preparative PL-SAX 4000 Å 30 µm 50×150 mm 1 VARIPL1751-3703
Preparative PL-SAX 1000 Å 30 µm 25×150 mm 1 VARIPL1251-3702
Preparative PL-SAX 1000 Å 30 µm 100×300 mm 1 VARIPL1851-3102
Preparative PL-SAX 1000 Å 10 µm 25×150 mm 1 VARIPL1251-3102
Preparative PL-SAX 1000 Å 10 µm 50×150 mm 1 VARIPL1751-3102
Preparative PL-SAX 1000 Å 10 µm 100×300 mm 1 VARIPL1851-2102
Preparative PL-SAX 4000 Å 30 µm 100×300 mm 1 VARIPL1851-3103
Preparative PL-SAX 4000 Å 10 µm 100×300 mm 1 VARIPL1851-2103
PL-SCX
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical - 1000 Å 8 µm 7,5×50 mm 1 VARIPL1145-1802
Analytical - 1000 Å 10 µm 4,6×150 mm 1 VARIPL1545-3102
Analytical - 1000 Å 10 µm 4,6×250 mm 1 VARIPL1545-5102
Analytical - 1000 Å 30 µm 4,6×150 mm 1 VARIPL1545-3702
Analytical - 1000 Å 30 µm 4,6×250 mm 1 VARIPL1545-5702
Analytical - 4000 Å 8 µm 7,5×50 mm 1 VARIPL1145-1803
Analytical - 4000 Å 10 µm 4,6×150 mm 1 VARIPL1545-3103
Analytical - 4000 Å 10 µm 4,6×250 mm 1 VARIPL1545-5103
Analytical - 4000 Å 30 µm 4,6×150 mm 1 VARIPL1545-3703
Analytical - 4000 Å 30 µm 4,6×250 mm 1 VARIPL1545-5703
Preparative PL-SCX 1000 Å 10 µm 50×150 mm 1 VARIPL1745-3102
Preparative PL-SCX 4000 Å 10 µm 100×300 mm 1 VARIPL1845-2103
Preparative PL-SCX 4000 Å 30 µm 50×150 mm 1 VARIPL1745-3703
Preparative PL-SCX 1000 Å 10 µm 25×150 mm 1 VARIPL1245-3102
Preparative PL-SCX 4000 Å 10 µm 25×50 mm 1 VARIPL1245-1103
Preparative PL-SCX 1000 Å 10 µm 100×300 mm 1 VARIPL1845-2102
Preparative PL-SCX 1000 Å 30 µm 50×150 mm 1 VARIPL1745-3702
Preparative PL-SCX 4000 Å 10 µm 50×150 mm 1 VARIPL1745-3103
Preparative PL-SCX 1000 Å 10 µm 25×50 mm 1 VARIPL1245-1102
Preparative PL-SCX 1000 Å 30 µm 100×300 mm 1 VARIPL1845-3102
Preparative PL-SCX 4000 Å 30 µm 25×150 mm 1 VARIPL1245-3703
Preparative PL-SCX 4000 Å 10 µm 25×150 mm 1 VARIPL1245-3103
Preparative PL-SCX 4000 Å 30 µm 100×300 mm 1 VARIPL1845-3103
Preparative PL-SCX 1000 Å 30 µm 25×150 mm 1 VARIPL1245-3702
Bulk media
Opis Particle size Veľkosť pórov Pk Číslo v katalógu VWR Množstvo
Bulk media, PL-SAX, 100 kg 30 µm 4000 Å 1 VARIPL1451-4703
Bulk media, PL-SAX, 100 kg 30 µm 1000 Å 1 VARIPL1451-4702
Bulk media, PL-SAX, 100 kg 10 µm 4000 Å 1 VARIPL1451-4103
Bulk media, PL-SAX, 100 kg 10 µm 1000 Å 1 VARIPL1451-4102
Bulk media, PL-SAX, 10 g 30 µm 4000 Å 1 VARIPL1451-2703
Bulk media, PL-SAX, 10 g 30 µm 1000 Å 1 VARIPL1451-2702
Bulk media, PL-SAX, 10 g 10 µm 4000 Å 1 VARIPL1451-2103
Bulk media, PL-SAX, 10 g 10 µm 1000 Å 1 VARIPL1451-2102
Bulk media, PL-SAX, 1 kg 30 µm 4000 Å 1 VARIPL1451-6703
Bulk media, PL-SAX, 1 kg 30 µm 1000 Å 1 VARIPL1451-6702
Bulk media, PL-SAX, 1 kg 10 µm 4000 Å 1 VARIPL1451-6103
Bulk media, PL-SAX, 1 kg 10 µm 1000 Å 1 VARIPL1451-6102
Bulk media, PL-SCX, 100 kg 30 µm 1000 Å 1 VARIPL1445-4702
Bulk media, PL-SCX, 100 kg 10 µm 4000 Å 1 VARIPL1445-4103
Bulk media, PL-SCX, 100 kg 10 µm 1000 Å 1 VARIPL1445-4102
Bulk media, PL-SCX, 100 kg 30 µm 4000 Å 1 VARIPL1445-4703
Bulk media, PL-SCX, 1 kg 10 µm 4000 Å 1 VARIPL1445-6103
Bulk media, PL-SCX, 1 kg 10 µm 1000 Å 1 VARIPL1445-6102
Bulk media, PL-SCX, 1 kg 30 µm 4000 Å 1 VARIPL1445-6703
Bulk media, PL-SCX, 1 kg 30 µm 1000 Å 1 VARIPL1445-6702
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo