Chcete sa opäť prihlásiť?
Laboratory syringes, two ring, Dosys™ 160 series
Dodávateľ: SOCOREX ISBA
Dosys™
Laboratory syringes are intended for safe and reliable liquid distribution, the selection of pistol-grip and two-ring instruments comes in a wide volume range.
  • Optimal hand balance and ergonomics
  • Fast and reliable volume setting
  • Plunger tightness without O-ring
  • High chemical resistance
Dosys™ 162 basic syringes are fill-by-fill (automatic) through needle or cannula, with luer lock nozzle.
Dosys™ 163 classic syringes are self-refilling automatic, with integrated valve system and luer lock nozzle.
Dosys™ 164 premium syringes are self-refilling automatic, with detachable valve system, enhanced corrosion resistance and luer lock nozzle.
Informace o dodávce: Dosys™ 162 basic package includes syringe, PVC protection sleeve and operating instructions. Dosys™ 163 classic package includes syringe, one-meter silicone feed tube, sinker, PVC protection sleeve, aspiration and vent cannulas, set of spares for valve, operating instructions. Dosys™ 164 premium package includes syringe, one-meter silicone feed tube, sinker, PVC protection sleeve, aspiration, vent and dosing cannulas, set of spares for valve, operating instructions.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Dosys™ 163 classic
Kapacita Graduation Pk Číslo v katalógu VWR Množstvo
0,025 - 0,3 ml 0,025 ml 1 SOCO163.05003
0,1 - 0,5 ml 0,1 ml 1 SOCO163.05005
0,1 - 1 ml 0,05 ml 1 SOCO163.0501
0,3 - 2 ml 0,1 ml 1 SOCO163.0502
0,5 - 5 ml 0,5 ml 1 SOCO163.0505
1 - 10 ml 1 ml 1 SOCO163.0510
Dosys™ 164 premium
Kapacita Graduation Pk Číslo v katalógu VWR Množstvo
0,1 - 1 ml 0,05 ml 1 SOCO164.0501
0,3 - 2 ml 0,1 ml 1 SOCO164.0502
0,5 - 5 ml 0,5 ml 1 SOCO164.0505
1 - 10 ml 1,0 ml 1 SOCO164.0510
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo