Chcete sa opäť prihlásiť?
HPLC columns, biochemical analysis, NUCLEOGEL® RP
Dodávateľ: MACHEREY-NAGEL
NUCLEOGEL®
Polymer based reversed phase columns for biochemical applications.
  • Polystyrene resin cross-linked with divinylbenzene
  • Reversed phase application under extreme pH (pH stability 1 - 13)
  • Eluent in column acetonitrile/water
NUCLEOGEL® RP columns with particle size of 100 Å are applied for reversed phase separations of small molecules such as pharmaceuticals with basic properties, nucleosides and nucleotides up to 5000 daltons; columns with 300 Å for large biomolecules (proteins, peptides etc.).
The Valco type guard column cartridge is used for analytical Valco column with the guard column holder B (MANA719539).
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical RP 100 Å 5 µm 4,6×150 mm 1 MANA719454
Analytical RP 100 Å 5 µm 4,6×250 mm 1 MANA719455
Analytical RP 100 Å 8 µm 4,6×150 mm 1 MANA719456
Analytical RP 100 Å 8 µm 4,6×250 mm 1 MANA719520
Analytical RP 300 Å 5 µm 4,6×50 mm 1 MANA719459
Analytical RP 300 Å 8 µm 4,6×50 mm 1 MANA719460
Guard columns
Column format Phase Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column RP 3,0×5 mm 2 MANA719542
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Max. pressure 180 bar
pH range 1 - 13
USP code L21
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo