Chcete sa opäť prihlásiť?
Reversed phase HPLC columns, biochemical analysis, NUCLEOSIL® MPN
Dodávateľ: MACHEREY-NAGEL
NUCLEOSIL®
Silica based reversed phase materials with monomerically bonded alkyl chains for RP chromatography that is ideal for seperation of biological macromolecules.
  • Based on robust spherical silica gel
  • Predominantly hydrophobic forces with a small portion of hydrophilic interactions
  • Eluent in column methanol
NUCLEOSIL® C18 MPN columns with particle size of 100 Å are suited for small hydrophobic peptides; C4 MPN with 300 Å for larger hydrophobic peptides and very different proteins.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C18 MPN 100 Å 5 µm 4,0×125 mm 1 MANA720230.40
Analytical C18 MPN 100 Å 5 µm 4,0×250 mm 1 MANA720231.40
Analytical C18 MPN 120 Å 3 µm 4,0×125 mm 1 MANA720232.40
Analytical C4 MPN 300 Å 5 µm 4,0×50 mm 1 MANA720244.40
Analytical C4 MPN 300 Å 5 µm 4,0×125 mm 1 MANA720045.40
Analytical C4 MPN 300 Å 5 µm 4,0×250 mm 1 MANA720245.40
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Description C4 MPN C18 MPN C4 MPN
Max. pressure 250 bar
pH range 2 – 8
USP code L26 L1 L26
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo