Chcete sa opäť prihlásiť?
HPLC columns, NUCLEOSIL® C18 Nautilus
Dodávateľ: MACHEREY-NAGEL
NUCLEOSIL®
NUCLEOSIL® C18 Nautilus is an endcapped octadecyl modified silica phase with an embedded polar group, using for HPLC.
  • Robust spherical silica gel
  • Hydrophobic phase with polar properties that is suited for 100% aqueous eluents
  • Separation mechanism based on hydrophobic and polar interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C18 Nautilus 100 Å 3 µm 4,0×125 mm 1 MANA720472.40
Analytical C18 Nautilus 100 Å 3 µm 4,6×125 mm 1 MANA720472.46
Analytical C18 Nautilus 100 Å 3 µm 4,6×150 mm 1 MANA720471.46
Analytical C18 Nautilus 100 Å 5 µm 4,0×125 mm 1 MANA720430.40
Analytical C18 Nautilus 100 Å 5 µm 4,0×250 mm 1 MANA720431.40
Analytical C18 Nautilus 100 Å 5 µm 4,6×125 mm 1 MANA720430.46
Analytical C18 Nautilus 100 Å 5 µm 4,6×150 mm 1 MANA720432.46
Analytical C18 Nautilus 100 Å 5 µm 4,6×250 mm 1 MANA720431.46
Guard columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column C18 Nautilus 100 Å 3 µm 3,0×4 mm 3 MANA721649.30
Guard column C18 Nautilus 100 Å 5 µm 3,0×4 mm 3 MANA721133.30
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Endcapped
USP code
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo