Chcete sa opäť prihlásiť?
Capillary columns, Select Biodiesel
Dodávateľ: Agilent Technologies
Select Biodiesel columns address the key challenge of good column lifetime when operating at very high temperatures up to 400 °C.
  • Complete set of biodiesel columns for full compliance and ease-of-use
  • UltiMetal technology provides high accuracy and longevity
  • Designed and pre-tested for complete confidence in results
  • Results are more consistent
  • No column breakage
Select Biodiesel columns are offered with a pre-coupled retention gap that is leak tested prior to shipment, making life much easier for the operator. This short piece of tubing not only enhances the analytical separation but also dramatically simplifies automation when using the column with a column inlet as specified in the standard methods.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Column format Phase Film thickness Int.Ø×L Pk Číslo v katalógu VWR Množstvo
UltiMetal retention gap, methyl deactivated Capillary Select Biodiesel - 0,53 mm × 2 m 1 VARICP6530
For gylcerides, UltiMetal, with retention gap Capillary Select Biodiesel 0,10 µm 0,32 mm × 10 m 1 VARICP9076
For gylcerides, UltiMetal Capillary Select Biodiesel 0,10 µm 0,32 mm × 10 m 1 VARICP9077
For gylcerides, UltiMetal, with retention gap Capillary Select Biodiesel 0,10 µm 0,32 mm × 15 m 1 VARICP9078
For gylcerides, UltiMetal Capillary Select Biodiesel 0,10 µm 0,32 mm × 15 m 1 VARICP9079
For FAME, fused silica Capillary Select Biodiesel 0,25 µm 0,32 mm × 30 m 1 VARICP9080
For Methanol, fused silica Capillary Select Biodiesel 3,00 µm 0,32 mm × 30 m 1 VARICP9083
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo