Chcete sa opäť prihlásiť?
Bottles, wide neck, with NS stopper
Dodávateľ: VITLAB
Made from opaque, pigmented PP, with NS stopper made of PP.
  • Good chemical resistance
  • Ideal for long-term storage of liquids
  • Autoclavable at 121 °C (2 bar) according to DIN EN 285
For light-sensitive substances. Wider mouth for simple filling, also ideal for powders and paste-like materials.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita NS Vonkajší Ø. Výška Pk Číslo v katalógu VWR Množstvo
1000 ml 60/46 108 mm 214 mm 10 VITL1018940
500 ml 45/40 87 mm 183 mm 10 VITL1017940
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo