Chcete sa opäť prihlásiť?
PLOT columns, CP-PoraBOND Q
Dodávateľ: Agilent Technologies
PLOT columns are ideal for separating compounds that are gases at room temperatures.
  • For analysing volatile solvents and hydrocarbons
  • Bonded PLOT column for more reliable results
  • Extended analysis of hydrocarbons for broader application range
  • Increased maximum temperature for greater productivity
  • Porous polymer is pure and has virtually no catalytic activity
The use of bonding technology in the CP-PoraBOND Q also reduces the presence of loose particles that cause detector spiking, so there is no need for particle traps.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Column format Film thickness Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Capillary 3,00 µm 0,25 mm × 10 m 1 VARICP7347
Capillary 5,00 µm 0,32 mm × 10 m 1 VARICP7350
Capillary 5,00 µm 0,32 mm × 10 m 1 VARICP7350I5
Capillary 10,00 µm 0,53 mm × 10 m 1 VARICP7353
Capillary 10,00 µm 0,53 mm × 10 m 1 VARICP7353I5
Capillary 3,00 µm 0,25 mm × 25 m 1 VARICP7348
Capillary 5,00 µm 0,32 mm × 25 m 1 VARICP7351
Capillary 5,00 µm 0,32 mm × 25 m 1 VARICP7351I5
Capillary 10,00 µm 0,53 mm × 25 m 1 VARICP7354
Capillary 10,00 µm 0,53 mm × 25 m 1 VARICP7354I5
Capillary 5,00 µm 0,32 mm × 50 m 1 VARICP7352
Capillary 5,00 µm 0,32 mm × 50 m 1 VARICP7352I5
Capillary 10,00 µm 0,53 mm × 50 m 1 VARICP7355
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo