Chcete sa opäť prihlásiť?
Bulb pipettes (one mark), BLAUBRAND®, class AS, USP
Dodávateľ: Brand
BLAUBRAND®
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • USP batch certificate available on request, also available with USP individual certificate
Certifikace: DIN EN ISO 648; DE-M marking.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Tolerancia Dĺžka Pk Číslo v katalógu VWR Množstvo
0,5 ml 0,005 ± ml 300 mm 12 612-4760
100 ml 0,08 ± ml 585 mm 6 612-4801
10 ml 0,02 ± ml 440 mm 6 612-4794
15 ml 0,03 ± ml 510 mm 6 612-4795
1 ml 0,006 ± ml 300 mm 12 612-4785
20 ml 0,03 ± ml 510 mm 6 612-4796
25 ml 0,03 ± ml 520 mm 6 612-4797
2 ml 0,006 ± ml 330 mm 12 612-4786
30 ml 0,03 ± ml 520 mm 6 612-4798
3 ml 0,01 ± ml 330 mm 6 612-4787
40 ml 0,05 ± ml 540 mm 6 612-4799
4 ml 0,01 ± ml 400 mm 6 612-4788
50 ml 0,05 ± ml 540 mm 6 612-4800
5 ml 0,01 ± ml 400 mm 6 612-4789
6 ml 0,01 ± ml 400 mm 6 612-4790
7 ml 0,01 ± ml 400 mm 6 612-4791
8 ml 0,02 ± ml 440 mm 6 612-4792
9 ml 0,02 ± ml 440 mm 6 612-4793
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo