Chcete sa opäť prihlásiť?
HPLC columns, Accucore™ Phenyl-X
Dodávateľ: Thermo Scientific
Accucore™
These columns offer a unique reversed-phase shape selectivity with high aromatic selectivity. Based on Core Enhanced Technology, they provide fast, high resolution separations without the elevated back pressures required by sub-2 µm particles.
  • Fast separations with high resolution
  • Low back pressure
  • Compatible with highly aqueous mobile phases
  • Low column bleed
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical Phenyl 80 Å 2,6 µm 4,6×150 mm 1 554-2314
Analytical Phenyl 80 Å 2,6 µm 2,1×100 mm 1 554-2309
Analytical Phenyl 80 Å 2,6 µm 2,1×250 mm 1 554-2315
Analytical Phenyl 80 Å 2,6 µm 3,0×100 mm 1 554-2310
Analytical Phenyl 80 Å 2,6 µm 2,1×50 mm 1 554-2306
Analytical Phenyl 80 Å 2,6 µm 3,0×50 mm 1 554-2307
Analytical Phenyl 80 Å 2,6 µm 2,1×150 mm 1 554-2312
Analytical Phenyl 80 Å 2,6 µm 3,0×150 mm 1 554-2313
Analytical Phenyl 80 Å 2,6 µm 4,6×100 mm 1 554-2311
Accucore™ defender guard columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard cartridge Phenyl 80 Å 2,6 µm 2,1×10 mm 4 554-2348
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo