Chcete sa opäť prihlásiť?
Isolator gloves, CSM
Dodávateľ: Piercan
These gloves made of chlorosulphonated polyethylene provide excellent resistance to chemicals, sterilisation agents (hydrogen peroxide) and sterilisation processes (gamma and beta radiation).
  • High resistance to ozone and UV
  • Excellent flexibility and dexterity
  • Excellent behavior to ionising radiations
Ideal for life science applications (pharmaceutical / medical).
EN 388 (4XX1), EN 374 (AKL), EN 374-2003, EN 421; Cat. III
Certifikace: EN 388 (4XX1), EN 374 (AKL), EN 374-2003, EN 421; Cat. III
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Model Glove size Farba Dĺžka Hrúbka Ø Pk Číslo v katalógu VWR Množstvo
Ambidextrous 7 White 330 mm 0,4 mm 110 mm 1 112-5286
Ambidextrous 8 White 330 mm 0,4 mm 110 mm 1 112-5287
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo