Chcete sa opäť prihlásiť?
HPLC columns, NUCLEOSIL® C6H5
Dodávateľ: MACHEREY-NAGEL
NUCLEOSIL®
NUCLEOSIL® C6H5 is a non-endcapped phenyl modified silica phase for HPLC.
  • Robust spherical silica gel
  • Weakly hydrophobic, non-endcapped phase with aromatic selectivity for RP chromatography
  • Separation mechanism is especially based on π–π interactions as well as on hydrophobic and some residual silanol interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C6H5 100 Å 5 µm 4,6×250 mm 1 MANA720956.46
Analytical C6H5 100 Å 7 µm 4,0×250 mm 1 MANA720019.40
Analytical C6H5 100 Å 7 µm 4,6×250 mm 1 MANA720019.46
Guard columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column C6H5 100 Å 5 µm 3,0×4 mm 3 MANA721137.30
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Endcapped
USP code
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo