Chcete sa opäť prihlásiť?
Beakers, low form
Dodávateľ: VITLAB
PTFE, with spout.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +260 °C
  • Flat bottom for good heat transfer, for example when used on regulated hotplates
Thick walled, with reinforced lip and exceptionally smooth inner walls to prevent contamination. Non stick. Low gas permeability.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Výška Ext.Ø Farba Pk Číslo v katalógu VWR Množstvo
5 ml 24 mm 22 mm Opaque/white 1 VITL112197
10 ml 39 mm 25 mm Opaque/white 1 VITL112297
25 ml 47 mm 32 mm Opaque/white 1 VITL112397
50 ml 55 mm 42 mm Opaque/white 1 VITL112497
100 ml 68 mm 55 mm Opaque/white 1 VITL112597
250 ml 93 mm 62 mm Opaque/white 1 VITL112697
500 ml 126 mm 81 mm Opaque/white 1 VITL112797
1000 ml 157 mm 102 mm Opaque/white 1 VITL112897
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo