Chcete sa opäť prihlásiť?
PCR tubes, 0,1 ml, with flat cap
Dodávateľ: Thermo Scientific
ABgene®
PP ultra-thin walled PCR tubes. Thinner walls offer less thermal resistance for transferring heat from the block to the sample within the tube. This results in the ability to perform very rapid protocols by reducing incubation times during PCR.
PP, clear or white
Ultra-thin walled PCR tubes.
  • Caps form a secure seal, yet are easy to open and close
  • Compatible with thermal cyclers accepting low-profile 0,2 ml tubes
  • Ultra-thin wall (UTW) low profile for fast PCR applications
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Farba Kapacita Balené Pk Číslo v katalógu VWR Množstvo
Flat capped tubes, ultra-thin walled Clear 0,1 ml 960 tubes 960 731-0405
Flat capped tubes, ultra-thin walled White 0,1 ml 960 tubes 960 731-0406
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo