Chcete sa opäť prihlásiť?
Dropping bottles
Dodávateľ: VITLAB
LDPE, translucent.
  • With dropper insert and HDPE screw cap
  • Used for food safe products
Transparent narrow mouth bottles with extra-long, fine dropping tip for accurate dispensing.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Výška Vonkajší Ø. Závit Pk Číslo v katalógu VWR Množstvo
50 ml 129 mm 37 mm GL 18 24 VITL94587
100 ml 155 mm 43 mm GL 18 24 VITL94687
250 ml 183 mm 65 mm GL 25 12 VITL94987
500 ml 228 mm 74 mm GL 25 12 VITL95087
1000 ml 269 mm 92 mm GL 32 12 VITL95187
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo