Chcete sa opäť prihlásiť?
Bottles, narrow neck, LDPE, with screw cap
Dodávateľ: VITLAB
LDPE, translucent, with PP screw cap.
  • Flexible material with good resilience
  • Food safe
  • FDA conform
With high shoulders.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Závit Vonkajší Ø. Výška Pk Číslo v katalógu VWR Množstvo
1000 ml GL 32 92 mm 221 mm 12 VITL95189
100 ml GL 18 43 mm 95 mm 24 VITL94689
250 ml GL 25 58 mm 135 mm 12 VITL94989
500 ml GL 25 74 mm 180 mm 12 VITL95089
50 ml GL 18 37 mm 73 mm 24 VITL94589
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo