Chcete sa opäť prihlásiť?
Full face masks against gas, vapour and dust; 9000 series
Dodávateľ: Moldex
Extremely lightweight, reusable full face mask (only 360 g) made from soft, TPE material.
  • Frameless panoramic lens
  • Easy access and easy to remove
  • Easy maintenance and cleaning, no tools required
  • With EasyLock® filter system (no additional adapters or parts necessary) or with EN 148-1 connectors
Certifikace: EN 136:1998, CL2
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Typ Veľkosť Pk Číslo v katalógu VWR Množstvo
Mask body, TPE, with EasyLock® connectors S 1 111-0709
Mask body, TPE, with EasyLock® connectors M 1 111-0708
Mask body, TPE, with EasyLock® connectors L 1 111-0707
Mask body, TPE, with EN 148-1 connectors S 1 111-0706
Mask body, TPE, with EN 148-1 connectors M 1 111-0705
Mask body, TPE, with EN 148-1 connectors L 1 111-0704
Mask body, TPE, incl. 2× A1B1E1K1P3 R filters mounted to mask M 1 KIT 111-1188
Filters for Masks 7000 and 9000 Series, EasyLock®
Opis Typ Trieda Pk Číslo v katalógu VWR Množstvo
9010, particle filter 9010, particle filter P1R 20 111-1180
9020, particle filter 9020, particle filter P2 20 111-0703
9022, particle filter + ozone 9022, particle filter + ozone P2 12 111-1181
9030, particle filter 9030, particle filter P3 12 111-0702
9032, particle filter + ozone 9032, particle filter + ozone P3 12 111-1182
9100, gas filter 9100, gas filter A1 10 111-0701
9120, pre-assembled filter 9120, pre-assembled filter A1P2R 8 111-1183
9200, gas filter 9200, gas filter A2 8 111-1184
9230, pre-assembled filter 9230, pre-assembled filter A2P3 6 111-1185
9300, gas filter 9300, gas filter A1B1E 10 111-1186
9400, gas filter 9400, gas filter A1B1E1K1 10 111-0700
9430, pre-assembled filter 9430, pre-assembled filter A1B1E1K1P3 6 111-1187
9500, gas filter 9500, gas filter A2B2E1 8 111-1189
9600, gas filter 9600, gas filter AX 8 111-0699
9730, pre-assembled filters 9730, pre-assembled filters A1B1E1K1HGP3 6 111-1190
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo