Chcete sa opäť prihlásiť?
Accurate digital refactometers, RX-i
Dodávateľ: Atago
Accurate digital refractometers featuring intuitive colour touch screen technology makes the RX-i range easy to use and navigate around the operating modes.
  • USB flash drive and self diagnosis capability
  • Peltier thermo-module to control temperature
  • Measurement starts after the sample reaches the target temperature
  • Refractive index and Brix at the target temperature are speedily displayed
  • Temperature control from 5 to 75 °C allows for measurement of RI or °Brix for a wide range of samples including emulsions
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Model Rozsah Index lomu Rozlíšenie Pk Číslo v katalógu VWR Množstvo
RX5000i 0,00 − 100,00 °B 1,32422 − 1,58000  nD 0,03 °B ; 0,00004 nD 1 ATAG3276
RX-5000i-Plus 0,00 − 100,00 °Bx 1,32422 − 1,58000  nD 0,005 °Bx ; 0,00001 nD 1 ATAG3275
RX7000i 0,00 − 100,00 °B 1,32422 − 1,70000  nD 0,01 °B ; 0,0001 nD 1 ATAG3279
RX9000i 0,00 − 100,00 °B 1,32422 − 1,70000  nD 0,01 °B ; 0,0001 nD 1 ATAG3278
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Temperature stability ±0,05 °C
Temperature measurement 5...75 °C
Prism Artificial sapphire
Displej Colour touch screen
W×D×H 370×260×140 mm
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo