Chcete sa opäť prihlásiť?
HPLC columns, HarmonySecure™
Dodávateľ: ES INDUSTRIES
HarmonySecure™
These columns are equivalent to Waters® Symmetry™ Shield columns. They utilise polar embedded technology for superior HPLC analysis.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
HarmonySecure™ RP18
Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
RP18 100 Å 3,5 µm 3,0×100 mm 1 ESIN123121-HRSRP18
RP18 100 Å 3,5 µm 4,6×75 mm 1 ESIN195121-HRSRP18
RP18 100 Å 5 µm 2,1×100 mm 1 ESIN122221-HRSRP18
RP18 100 Å 5 µm 3,9×150 mm 1 ESIN13E221-HRSRP18
RP18 100 Å 5 µm 2,1×150 mm 1 ESIN132221-HRSRP18
RP18 100 Å 5 µm 3,0×250 mm 1 ESIN153221-HRSRP18
RP18 100 Å 3,5 µm 4,6×100 mm 1 ESIN125121-HRSRP18
RP18 100 Å 5 µm 4,6×100 mm 1 ESIN125221-HRSRP18
RP18 100 Å 3,5 µm 3,0×50 mm 1 ESIN113121-HRSRP18
RP18 100 Å 5 µm 4,6×150 mm 1 ESIN135221-HRSRP18
RP18 100 Å 3,5 µm 2,1×50 mm 1 ESIN112121-HRSRP18
RP18 100 Å 3,5 µm 2,1×100 mm 1 ESIN122121-HRSRP18
RP18 100 Å 5 µm 4,6×250 mm 1 ESIN155221-HRSRP18
RP18 100 Å 3,5 µm 4,6×150 mm 1 ESIN135121-HRSRP18
RP18 100 Å 5 µm 3,0×150 mm 1 ESIN133221-HRSRP18
RP18 100 Å 5 µm 2,1×50 mm 1 ESIN112221-HRSRP18
RP18 100 Å 3,5 µm 4,6×50 mm 1 ESIN115121-HRSRP18
RP18 100 Å 3,5 µm 2,1×150 mm 1 ESIN132121-HRSRP18
RP18 100 Å 3,5 µm 3,0×150 mm 1 ESIN133121-HRSRP18
RP18 100 Å 5 µm 4,6×50 mm 1 ESIN115221-HRSRP18
HarmonySecure™ RP8
Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
RP8 100 Å 5 µm 4,6×250 mm 1 ESIN155221-HRSRP8
RP8 100 Å 3,5 µm 3,0×150 mm 1 ESIN133121-HRSRP8
RP8 100 Å 3,5 µm 4,6×50 mm 1 ESIN115121-HRSRP8
RP8 100 Å 5 µm 3,0×150 mm 1 ESIN133221-HRSRP8
RP8 100 Å 3,5 µm 4,6×150 mm 1 ESIN135121-HRSRP8
RP8 100 Å 5 µm 2,1×150 mm 1 ESIN132221-HRSRP8
RP8 100 Å 3,5 µm 3,0×50 mm 1 ESIN113121-HRSRP8
RP8 100 Å 3,5 µm 2,1×150 mm 1 ESIN132121-HRSRP8
RP8 100 Å 3,5 µm 2,1×50 mm 1 ESIN112121-HRSRP8
RP8 100 Å 5 µm 3,0×250 mm 1 ESIN153221-HRSRP8
RP8 100 Å 5 µm 4,6×100 mm 1 ESIN125221-HRSRP8
RP8 100 Å 3,5 µm 4,6×100 mm 1 ESIN125121-HRSRP8
RP8 100 Å 5 µm 2,1×50 mm 1 ESIN112221-HRSRP8
RP8 100 Å 5 µm 4,6×50 mm 1 ESIN115221-HRSRP8
RP8 100 Å 3,5 µm 4,6×75 mm 1 ESIN195121-HRSRP8
RP8 100 Å 3,5 µm 3,0×100 mm 1 ESIN123121-HRSRP8
RP8 100 Å 5 µm 2,1×100 mm 1 ESIN122221-HRSRP8
RP8 100 Å 5 µm 3,9×150 mm 1 ESIN13E221-HRSRP8
RP8 100 Å 3,5 µm 2,1×100 mm 1 ESIN122121-HRSRP8
RP8 100 Å 5 µm 4,6×150 mm 1 ESIN135221-HRSRP8
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo