Chcete sa opäť prihlásiť?
SPE columns, CHROMABOND® SA
Dodávateľ: MACHEREY-NAGEL
CHROMABOND®
CHROMABOND® SA, Benzenesulfonic acid modified silica cation exchanger for SPE (SCX).
  • Base material silica, pH stability 2 - 8
  • Strongly acidic cation exchanger (capacity ~ 0,5 meq/g)
  • Adsorbent with hydrophobic and π-π interactions (benzene ring)
  • Ion exchange of organic compounds from aqueous matrix
  • Elution of interesting compounds with solvent systems, which compensate the ionic and nonpolar interactions
Recommended for amino acids, amines, chlorophyll and PCBs.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
CHROMABOND® SA polypropylene columns
Phase Column hardware Veľkosť pórov Particle size Kapacita Bed weight Pk Číslo v katalógu VWR Množstvo
SA PP 60 Å 45 µm 1 ml 100 mg 100 MANA730076
SA PP 60 Å 45 µm 3 ml 200 mg 50 MANA730275
SA PP 60 Å 45 µm 3 ml 500 mg 50 MANA730077
SA PP 60 Å 45 µm 3 ml 500 mg 50 MANA730077.250
SA PP 60 Å 45 µm 6 ml 500 mg 30 MANA730425
SA PP 60 Å 45 µm 6 ml 1000 mg 30 MANA730212
CHROMABOND® LV-SA
Phase Column hardware Veľkosť pórov Particle size Kapacita Bed weight Pk Číslo v katalógu VWR Množstvo
SA PP 60 Å 45 µm 15 ml 500 mg 30 MANA732083
CHROMABOND® MULTI 96 SA
Phase Column hardware Veľkosť pórov Particle size Kapacita Bed weight Pk Číslo v katalógu VWR Množstvo
SA PP 60 Å 45 µm 96×100 mg 1 KIT MANA738141.100M
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Surface area 500 m²/g
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo