Chcete sa opäť prihlásiť?
HPLC columns, HyperSelect™ HiPurity
Dodávateľ: ES INDUSTRIES
HyperSelect™
These columns are an alternative to the Thermo Fisher Hypersil™ columns.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
HyperSelect™ HiPurity C18
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C18 200 Å 3 µm 4×150 mm 1 ESIN1341A1HPCHIC18
Analytical C18 200 Å 3 µm 4×100 mm 1 ESIN1241A1HPCHIC18
Analytical C18 200 Å 3 µm 2,1×50 mm 1 ESIN1121A1HPCHIC18
Analytical C18 200 Å 3 µm 4,6×100 mm 1 ESIN1251A1HPCHIC18
Analytical C18 200 Å 5 µm 4,6×250 mm 1 ESIN1552A1HPCHIC18
Analytical C18 200 Å 5 µm 4×250 mm 1 ESIN1542A1HPCHIC18
Analytical C18 200 Å 5 µm 4×150 mm 1 ESIN1342A1HPCHIC18
Analytical C18 200 Å 5 µm 3,2×150 mm 1 ESIN13D2A1HPCHIC18
Analytical C18 200 Å 3 µm 3,2×50 mm 1 ESIN11D1A1HPCHIC18
Analytical C18 200 Å 5 µm 2,1×150 mm 1 ESIN1322A1HPCHIC18
Analytical C18 200 Å 5 µm 2,1×100 mm 1 ESIN1222A1HPCHIC18
Analytical C18 200 Å 3 µm 4,6×50 mm 1 ESIN1151A1HPCHIC18
Analytical C18 200 Å 3 µm 4×50 mm 1 ESIN1141A1HPCHIC18
Analytical C18 200 Å 5 µm 3,2×50 mm 1 ESIN11D2A1HPCHIC18
Analytical C18 200 Å 5 µm 4,6×150 mm 1 ESIN1352A1HPCHIC18
Analytical C18 200 Å 5 µm 4,6×100 mm 1 ESIN1252A1HPCHIC18
Analytical C18 200 Å 5 µm 4×50 mm 1 ESIN1142A1HPCHIC18
Analytical C18 200 Å 3 µm 3,2×250 mm 1 ESIN15D1A1HPCHIC18
Analytical C18 200 Å 5 µm 3,2×250 mm 1 ESIN15D2A1HPCHIC18
Analytical C18 200 Å 3 µm 4×250 mm 1 ESIN1541A1HPCHIC18
Analytical C18 200 Å 5 µm 3,2×100 mm 1 ESIN12D2A1HPCHIC18
Analytical C18 200 Å 3 µm 2,1×250 mm 1 ESIN1521A1HPCHIC18
Analytical C18 200 Å 5 µm 4×100 mm 1 ESIN1242A1HPCHIC18
Analytical C18 200 Å 5 µm 2,1×50 mm 1 ESIN1122A1HPCHIC18
Analytical C18 200 Å 3 µm 4,6×150 mm 1 ESIN1351A1HPCHIC18
Analytical C18 200 Å 5 µm 4,6×50 mm 1 ESIN1152A1HPCHIC18
Analytical C18 200 Å 3 µm 4,6×250 mm 1 ESIN1551A1HPCHIC18
Analytical C18 200 Å 3 µm 3,2×100 mm 1 ESIN12D1A1HPCHIC18
Analytical C18 200 Å 3 µm 2,1×150 mm 1 ESIN1321A1HPCHIC18
Analytical C18 200 Å 3 µm 2,1×100 mm 1 ESIN1221A1HPCHIC18
Analytical C18 200 Å 3 µm 3,2×150 mm 1 ESIN13D1A1HPCHIC18
Analytical C18 200 Å 5 µm 2,1×250 mm 1 ESIN1522A1HPCHIC18
HyperSelect™ HiPurity C8
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C8 200 Å 5 µm 4×50 mm 1 ESIN1142A1-HPCHIC8
Analytical C8 200 Å 5 µm 4×100 mm 1 ESIN1242A1-HPCHIC8
Analytical C8 200 Å 5 µm 4×150 mm 1 ESIN1342A1-HPCHIC8
Analytical C8 200 Å 5 µm 3,2×250 mm 1 ESIN15D2A1-HPCHIC8
Analytical C8 200 Å 5 µm 4,6×100 mm 1 ESIN1252A1-HPCHIC8
Analytical C8 200 Å 5 µm 2,1×50 mm 1 ESIN1122A1-HPCHIC8
Analytical C8 200 Å 5 µm 4,6×250 mm 1 ESIN1552A1-HPCHIC8
Analytical C8 200 Å 5 µm 4,6×50 mm 1 ESIN1152A1-HPCHIC8
Analytical C8 200 Å 5 µm 2,1×150 mm 1 ESIN1322A1-HPCHIC8
Analytical C8 200 Å 5 µm 2,1×100 mm 1 ESIN1222A1-HPCHIC8
Analytical C8 200 Å 5 µm 3,2×100 mm 1 ESIN12D2A1-HPCHIC8
Analytical C8 200 Å 5 µm 3,2×50 mm 1 ESIN11D2A1-HPCHIC8
Analytical C8 200 Å 5 µm 2,1×250 mm 1 ESIN1522A1-HPCHIC8
Analytical C8 200 Å 5 µm 3,2×150 mm 1 ESIN13D2A1-HPCHIC8
Analytical C8 200 Å 5 µm 4×250 mm 1 ESIN1542A1-HPCHIC8
Analytical C8 200 Å 5 µm 4,6×150 mm 1 ESIN1352A1-HPCHIC8
HyperSelect™ HiPurity C4
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C4 200 Å 5 µm 4,6×50 mm 1 ESIN1152A1-HPCHIC4
Analytical C4 200 Å 5 µm 4,6×100 mm 1 ESIN1252A1-HPCHIC4
Analytical C4 200 Å 5 µm 4×100 mm 1 ESIN1242A1-HPCHIC4
Analytical C4 200 Å 5 µm 3,2×50 mm 1 ESIN11D2A1-HPCHIC4
Analytical C4 200 Å 5 µm 4,6×150 mm 1 ESIN1352A1-HPCHIC4
Analytical C4 200 Å 5 µm 3,2×100 mm 1 ESIN12D2A1-HPCHIC4
Analytical C4 200 Å 5 µm 2,1×50 mm 1 ESIN1122A1-HPCHIC4
Analytical C4 200 Å 5 µm 4×250 mm 1 ESIN1542A1-HPCHIC4
Analytical C4 200 Å 5 µm 4×50 mm 1 ESIN1142A1-HPCHIC4
Analytical C4 200 Å 5 µm 2,1×150 mm 1 ESIN1322A1-HPCHIC4
Analytical C4 200 Å 5 µm 2,1×250 mm 1 ESIN1522A1-HPCHIC4
Analytical C4 200 Å 5 µm 4×150 mm 1 ESIN1342A1-HPCHIC4
Analytical C4 200 Å 5 µm 2,1×100 mm 1 ESIN1222A1-HPCHIC4
Analytical C4 200 Å 5 µm 3,2×150 mm 1 ESIN13D2A1-HPCHIC4
Analytical C4 200 Å 5 µm 3,2×250 mm 1 ESIN15D2A1-HPCHIC4
Analytical C4 200 Å 5 µm 4,6×250 mm 1 ESIN1552A1-HPCHIC4
HyperSelect™ HiPurity Cyano
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical Cyano 200 Å 5 µm 3,2×250 mm 1 ESIN15D2A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4,6×150 mm 1 ESIN1352A1-HPCHICN
Analytical Cyano 200 Å 5 µm 3,2×150 mm 1 ESIN13D2A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4,6×50 mm 1 ESIN1152A1-HPCHICN
Analytical Cyano 200 Å 5 µm 3,2×100 mm 1 ESIN12D2A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4×150 mm 1 ESIN1342A1-HPCHICN
Analytical Cyano 200 Å 5 µm 2,1×100 mm 1 ESIN1222A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4×250 mm 1 ESIN1542A1-HPCHICN
Analytical Cyano 200 Å 5 µm 2,1×150 mm 1 ESIN1322A1-HPCHICN
Analytical Cyano 200 Å 5 µm 3,2×50 mm 1 ESIN11D2A1-HPCHICN
Analytical Cyano 200 Å 5 µm 2,1×250 mm 1 ESIN1522A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4×50 mm 1 ESIN1142A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4×100 mm 1 ESIN1242A1-HPCHICN
Analytical Cyano 200 Å 5 µm 2,1×50 mm 1 ESIN1122A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4,6×100 mm 1 ESIN1252A1-HPCHICN
Analytical Cyano 200 Å 5 µm 4,6×250 mm 1 ESIN1552A1-HPCHICN
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo