Chcete sa opäť prihlásiť?
HPLC columns, HyperSelect™ BDS
Dodávateľ: ES INDUSTRIES
HyperSelect™
These columns are an alternative to the Thermo Fisher Hypersil™ columns.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
HyperSelect™ BDS C18
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical BDS C18 120 Å 5 µm 2,1×150 mm 1 ESIN132291HPBDSC18
Analytical BDS C18 120 Å 5 µm 2,1×100 mm 1 ESIN122291HPBDSC18
Analytical BDS C18 120 Å 3 µm 4,6×50 mm 1 ESIN115191HPBDSC18
Analytical BDS C18 120 Å 3 µm 3,2×250 mm 1 ESIN15D191HPBDSC18
Analytical BDS C18 120 Å 3 µm 4×250 mm 1 ESIN154191HPBDSC18
Analytical BDS C18 120 Å 3 µm 4×150 mm 1 ESIN134191HPBDSC18
Analytical BDS C18 120 Å 5 µm 3,2×100 mm 1 ESIN12D291HPBDSC18
Analytical BDS C18 120 Å 3 µm 3,2×150 mm 1 ESIN13D191HPBDSC18
Analytical BDS C18 120 Å 3 µm 3,2×50 mm 1 ESIN11D191HPBDSC18
Analytical BDS C18 120 Å 3 µm 2,1×50 mm 1 ESIN112191HPBDSC18
Analytical BDS C18 120 Å 3 µm 3,2×100 mm 1 ESIN12D191HPBDSC18
Analytical BDS C18 120 Å 5 µm 4×250 mm 1 ESIN154291HPBDSC18
Analytical BDS C18 120 Å 5 µm 3,2×50 mm 1 ESIN11D291HPBDSC18
Analytical BDS C18 120 Å 3 µm 4,6×100 mm 1 ESIN125191HPBDSC18
Analytical BDS C18 120 Å 5 µm 4×100 mm 1 ESIN124291HPBDSC18
Analytical BDS C18 120 Å 3 µm 4,6×250 mm 1 ESIN155191HPBDSC18
Analytical BDS C18 120 Å 5 µm 4,6×50 mm 1 ESIN115291HPBDSC18
Analytical BDS C18 120 Å 5 µm 4×150 mm 1 ESIN134291HPBDSC18
Analytical BDS C18 120 Å 5 µm 3,2×250 mm 1 ESIN15D291HPBDSC18
Analytical BDS C18 120 Å 5 µm 2,1×250 mm 1 ESIN152291HPBDSC18
Analytical BDS C18 120 Å 5 µm 4,6×250 mm 1 ESIN155291HPBDSC18
Analytical BDS C18 120 Å 3 µm 4,6×150 mm 1 ESIN135191HPBDSC18
Analytical BDS C18 120 Å 3 µm 4×50 mm 1 ESIN114191HPBDSC18
Analytical BDS C18 120 Å 5 µm 4×50 mm 1 ESIN114291HPBDSC18
Analytical BDS C18 120 Å 5 µm 2,1×50 mm 1 ESIN112291HPBDSC18
Analytical BDS C18 120 Å 3 µm 2,1×100 mm 1 ESIN122191HPBDSC18
Analytical BDS C18 120 Å 5 µm 4,6×150 mm 1 ESIN135291HPBDSC18
Analytical BDS C18 120 Å 5 µm 4,6×100 mm 1 ESIN125291HPBDSC18
Analytical BDS C18 120 Å 3 µm 2,1×150 mm 1 ESIN132191HPBDSC18
Analytical BDS C18 120 Å 3 µm 2,1×250 mm 1 ESIN152191HPBDSC18
Analytical BDS C18 120 Å 5 µm 3,2×150 mm 1 ESIN13D291HPBDSC18
Analytical BDS C18 120 Å 3 µm 4×100 mm 1 ESIN124191HPBDSC18
HyperSelect™ BDS C8
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical BDS C8 120 Å 5 µm 4×100 mm 1 ESIN124291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4,6×50 mm 1 ESIN115191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 3,2×250 mm 1 ESIN15D191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4×150 mm 1 ESIN134191HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4,6×50 mm 1 ESIN115291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 3,2×100 mm 1 ESIN12D291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 3,2×150 mm 1 ESIN13D291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4×150 mm 1 ESIN134291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 2,1×100 mm 1 ESIN122191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4×100 mm 1 ESIN124191HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4×250 mm 1 ESIN154291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 2,1×250 mm 1 ESIN152291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 2,1×150 mm 1 ESIN132291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4,6×100 mm 1 ESIN125291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4×50 mm 1 ESIN114291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4,6×100 mm 1 ESIN125191HPCBDSC8
Analytical BDS C8 120 Å 5 µm 3,2×250 mm 1 ESIN15D291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 2,1×100 mm 1 ESIN122291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4,6×250 mm 1 ESIN155291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4×250 mm 1 ESIN154191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4,6×150 mm 1 ESIN135191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4,6×250 mm 1 ESIN155191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 3,2×100 mm 1 ESIN12D191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 4×50 mm 1 ESIN114191HPCBDSC8
Analytical BDS C8 120 Å 5 µm 3,2×50 mm 1 ESIN11D291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 2,1×50 mm 1 ESIN112191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 3,2×150 mm 1 ESIN13D191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 2,1×150 mm 1 ESIN132191HPCBDSC8
Analytical BDS C8 120 Å 3 µm 2,1×250 mm 1 ESIN152191HPCBDSC8
Analytical BDS C8 120 Å 5 µm 4,6×150 mm 1 ESIN135291HPCBDSC8
Analytical BDS C8 120 Å 5 µm 2,1×50 mm 1 ESIN112291HPCBDSC8
Analytical BDS C8 120 Å 3 µm 3,2×50 mm 1 ESIN11D191HPCBDSC8
HyperSelect™ BDS Cyano
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical BDS cyano 120 Å 5 µm 3,2×250 mm 1 ESIN15D291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 2,1×150 mm 1 ESIN132191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 3,2×50 mm 1 ESIN11D291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 2,1×250 mm 1 ESIN152291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4×100 mm 1 ESIN124291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4×150 mm 1 ESIN134291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4×100 mm 1 ESIN124191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 3,2×50 mm 1 ESIN11D191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 2,1×50 mm 1 ESIN112191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 2,1×50 mm 1 ESIN112291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4×50 mm 1 ESIN114191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4,6×150 mm 1 ESIN135291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4,6×100 mm 1 ESIN125291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4×50 mm 1 ESIN114291HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 2,1×150 mm 1 ESIN132291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4×150 mm 1 ESIN134191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 3,2×100 mm 1 ESIN12D291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 3,2×250 mm 1 ESIN15D191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4×250 mm 1 ESIN154291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 2,1×100 mm 1 ESIN122191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4,6×150 mm 1 ESIN135191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4,6×250 mm 1 ESIN155291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4,6×250 mm 1 ESIN155191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 4,6×50 mm 1 ESIN115291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 2,1×250 mm 1 ESIN152191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 3,2×100 mm 1 ESIN12D191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4×250 mm 1 ESIN154191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 3,2×150 mm 1 ESIN13D291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 3,2×150 mm 1 ESIN13D191HPCBDSCN
Analytical BDS cyano 120 Å 5 µm 2,1×100 mm 1 ESIN122291HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4,6×50 mm 1 ESIN115191HPCBDSCN
Analytical BDS cyano 120 Å 3 µm 4,6×100 mm 1 ESIN125191HPCBDSCN
HyperSelect™ BDS Phenyl
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical BDS phenyl 120 Å 5 µm 3,2×50 mm 1 ESIN11D291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4,6×250 mm 1 ESIN155291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 3,2×150 mm 1 ESIN13D291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4,6×50 mm 1 ESIN115191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 3,2×250 mm 1 ESIN15D191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4,6×250 mm 1 ESIN155191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 2,1×50 mm 1 ESIN112291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 2,1×250 mm 1 ESIN152191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 2,1×100 mm 1 ESIN122191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 2,1×50 mm 1 ESIN112191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 2,1×250 mm 1 ESIN152291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4×50 mm 1 ESIN114191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4×150 mm 1 ESIN134191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 2,1×150 mm 1 ESIN132291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 2,1×150 mm 1 ESIN132191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4×250 mm 1 ESIN154291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 3,2×50 mm 1 ESIN11D191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4,6×150 mm 1 ESIN135191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4,6×100 mm 1 ESIN125191HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 3,2×150 mm 1 ESIN13D191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4,6×100 mm 1 ESIN125291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4,6×150 mm 1 ESIN135291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4,6×50 mm 1 ESIN115291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4×250 mm 1 ESIN154191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4×150 mm 1 ESIN134291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4×50 mm 1 ESIN114291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 3,2×250 mm 1 ESIN15D291HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 4×100 mm 1 ESIN124291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 4×100 mm 1 ESIN124191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 3,2×100 mm 1 ESIN12D291HPCBDSPH
Analytical BDS phenyl 120 Å 3 µm 3,2×100 mm 1 ESIN12D191HPCBDSPH
Analytical BDS phenyl 120 Å 5 µm 2,1×100 mm 1 ESIN122291HPCBDSPH
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo