Chcete sa opäť prihlásiť?
SPE columns, CHROMABOND® OH
Dodávateľ: MACHEREY-NAGEL
CHROMABOND®
CHROMABOND® OH, Diol modified silica phase for SPE.
  • Base material silica, pH stability 2 - 8
  • Carbon content 5,5%
  • Polar, properties similar to SiOH
Recommended for antibiotics and prostaglandins.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Phase Column hardware Veľkosť pórov Particle size Kapacita Bed weight Pk Číslo v katalógu VWR Množstvo
OH (Diol) PP 60 Å 45 µm 1 ml 100 mg 100 MANA730051
OH (Diol) PP 60 Å 45 µm 3 ml 200 mg 50 MANA730417
OH (Diol) PP 60 Å 45 µm 3 ml 500 mg 50 MANA730053
OH (Diol) PP 60 Å 45 µm 6 ml 500 mg 30 MANA730418
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
Carbon load 5,5%
Surface area 500 m²/g
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo