Chcete sa opäť prihlásiť?
Bottles, wide neck, with screw cap
Dodávateľ: VITLAB
LDPE, translucent, with screw cap made of PP.
  • Good chemical resistance
  • Good temperature resistance
  • Flexible material with good resilience
Wider mouth for simple filling, also ideal for powders and paste-like materials.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Kapacita Závit Vonkajší Ø. Výška Pk Číslo v katalógu VWR Množstvo
1000 ml GL 63 91 mm 226 mm 12 VITL94189
100 ml GL 32 47 mm 94 mm 24 VITL93489
250 ml GL 45 58 mm 146 mm 12 VITL93789
500 ml GL 45 76 mm 166 mm 12 VITL93989
50 ml GL 32 39 mm 87 mm 24 VITL93389
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo