Chcete sa opäť prihlásiť?
Accessories
Dodávateľ: MACHEREY-NAGEL
CHROMABOND®
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
Adapters (PE) for PP columns (15, 45 and 70 ml) 4 MANA730350.4
Adapters (PE) for PP columns (15, 45 and 70 ml) 10 MANA730385
Adapters (PE) for PP columns (30 and 70 ml) 1 KIT MANA730566
Adapters (PP) for PP columns (1, 3 and 6 ml) 4 MANA730100.4
Adapters (PP) for PP columns (1, 3 and 6 ml) 10 MANA730101
Adapters (PVDF) for glass columns (3 and 6 ml) 4 MANA730104.4
Adapters (PVDF) for glass columns (3 and 6 ml) 10 MANA730105
Glass fiber frits for glass columns, 3 ml 250 MANA730191
Glass fiber frits for glass columns, 6 ml 250 MANA730192
PE frits for LV PP columns 15 ml, for 100 mg adsorbent weight 250 MANA732019
PE frits for LV PP columns 15 ml, for 200/500 mg adsorbent weight 250 MANA732020
PE frits for PP columns, 1 ml 250 MANA730164
PE frits for PP columns, 3 ml 250 MANA730162
PE frits for PP columns, 6 ml 250 MANA730163
PE frits for PP columns, 15 ml 250 MANA730351
PE frits for PP columns, 30 ml 250 MANA730034
PE frits for PP columns, 45 ml 250 MANA730356
PE frits for PP columns, 70 ml 250 MANA730026
PE frits for PP columns, 150 ml 250 MANA730475
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo