Chcete sa opäť prihlásiť?
HPLC columns, PAH analysis, NUCLEOSIL® C18 PAH
Dodávateľ: MACHEREY-NAGEL
NUCLEOSIL®
Special octadecyl modified silica phase for HPLC analysis of PAHs.
  • Robust spherical silica gel
  • Polymeric octadecyl modification
  • Allows efficient gradient separation of the 16 PAH according to EPA
Detection of the separated PAHs by UV (250 to 280 nm) with diode array or with fluorescence detection at different wavelengths for excitation and emission. But acenaphthylene cannot be analyzed with fluorescence detection.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Analytical columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Analytical C18 PAH 100 Å 5 µm 2,0×250 mm 1 MANA720117.20
Analytical C18 PAH 100 Å 5 µm 3,0×150 mm 1 MANA720923.30
Analytical C18 PAH 100 Å 5 µm 3,0×250 mm 1 MANA720117.30
Analytical C18 PAH 100 Å 5 µm 4,0×150 mm 1 MANA720923.40
Analytical C18 PAH 100 Å 5 µm 4,0×250 mm 1 MANA720117.40
Analytical C18 PAH 100 Å 5 µm 4,6×250 mm 1 MANA720117.46
Guard columns
Column format Phase Veľkosť pórov Particle size Int.Ø×L Pk Číslo v katalógu VWR Množstvo
Guard column C18 PAH 100 Å 5 µm 2,0×4 mm 3 MANA721168.20
Guard column C18 PAH 100 Å 5 µm 3,0×4 mm 3 MANA721168.30
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
ŠPECIFIKÁCIE
USP code L1
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo