Chcete sa opäť prihlásiť?
Eco bins, SLK series
Dodávateľ: Multiroir
PP
  • Robust
  • Easy filling and emptying even when stacked
This system of stackable bins allows for quick and easy storage and identification.
Colour: Blue or red
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Š × H × V Pk Číslo v katalógu VWR Množstvo
100×215×75 mm 1 216-1131
150×230×125 mm 1 216-1133
150×315×125 mm 1 216-1137
210×350×150 mm 1 216-1141
315×500×200 mm 1 216-1143
100×215×75 mm 1 216-1065
150×230×125 mm 1 216-1132
150×315×125 mm 1 216-1135
210×350×150 mm 1 216-1139
315×500×200 mm 1 216-1142
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo