Chcete sa opäť prihlásiť?
Pallets, lightweight
Dodávateľ: Multiroir
HDPE, black
  • Models with skids or feet
  • Resistant to heat and cold
  • Easy cleaning
This pallets can be stacked one into the other to save space. They have a high load-bearing capacity, despite their low empty weight.
OBJEDNAŤ
Rozšíriť všetko / Schovať všetko
Opis Pk Číslo v katalógu VWR Množstvo
1000×1200 mm (D×W), 3 skids 1 216-1159
1000×1200 mm (D×W), 9 feet 1 216-1157
800×1200 mm (D×W), 3 skids 1 216-1144
800×1200 mm (D×W), 9 feet 1 216-1078
Vytvoriť čiarové kódy Formát čiarového kódu: 
Vytvoriť katalóg v PDF
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo