Keep my session open?
Ending In 
The session is expired
Vaša relácia uplynula. Pre Vašu bezpečnosť sme Vás odhlásili.
Chcete sa opäť prihlásiť?

Calcium phosphide, tech.

Dodávateľ: Alfa Aesar

CAS No.: 1305-99-3
Molecular Formula: Ca3P2
Formula Weight: 182.19
Physical Form: Lump
Density: 2.51
Sensitivity: Moisture Sensitive
Appearance: Red-brown
Melting Point: ca 1600°
Merck Reference: 14,1695
UN No.: 1360
MDL No.: MFCD00015986

Purification of copper and copper alloys, fire signals


Vzorec: Ca₃P₂
MW: 182,182 g/mol
MDL Number: MFCD00015986
Číslo CAS: 1305-99-3
EINECS: 215-142-0
UN: 1360
ADR: 4.3,I


 

OBJEDNÁVKA

Rozšíriť všetko / Schovať všetko
Pk Číslo v katalógu VWR Množstvo
1 kg 19115.A1
250 g 19115.30
Rozšíriť všetko / Schovať všetko
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ŠPECIFIKÁCIE

Density 2.51
Sensitivity Moisture Sensitive
Melting point ca 1600°
Formula weight 182.19
CAS No. 1305-99-3
Physical form Lump
Molecular formula Ca3P2
MDL No. MFCD00015986
UN No. 1360
Merck reference 14,1695
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter "x" if you don't know for VWR Chemicals.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Alebo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Popis Číslo v katalógu Availability Jednotka Vaša cena Price Per Množstvo