Tlačiť

Acetonitrile-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®

Dodávateľ: Merck


M-Clarity™ quality level = MQ200

SIAL485217-250MGEA 849.76 EUR
SIAL485217-250MG
Acetonitrile-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Acetonitrile-[¹³C₂]
Vzorec: C₂H₃N
Číslo MDL: MFCD00083867
Číslo CAS: 1722-25-4
OSN: 1648
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR