Tlačiť
DNA labelling system, Amersham Megaprime™
Dodávateľ: Cytiva
RPN1607EA 1359.49 EUR
GEHERPN1607
DNA labelling system, Amersham Megaprime™
Premixed, ambient-temperature-stable reactions with a color check for the labeling of DNA to specific activities of > 10⁹ dpm/µg.
  • For use with any radioactively labelled nucleotide
  • For labelling of DNA probes to specific activities >1×10⁹ dpm/µg in 10 minutes
  • No need to remove unincorporated nucleotides before hybridisation
  • Protocols for labelling directly from low melting point agarose
Features cloned Klenow fragment and random nonamers in the random-prime labelling technique.
Informace o dodávce: Each system includes the following reagents, sufficient for 30 or 60 reactions: Primer solution, labelling buffer, reaction buffer, enzyme, standard DNA solution, and carrier DNA solution.